Skip to Content
Merck

1019712

USP

Amiloride Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H7ClN4O2
CAS Number:
Molecular Weight:
202.60
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

amiloride

manufacturer/tradename

USP

mp

211-215 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COC(=O)c1nc(Cl)c(N)nc1N

InChI

1S/C6H7ClN4O2/c1-13-6(12)2-4(8)11-5(9)3(7)10-2/h1H3,(H4,8,9,11)

InChI key

KOOBYHRLTYIPTH-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amiloride Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Amiloride Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Amiloride Related Compound A


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library