Skip to Content
Merck
All Photos(1)

Key Documents

1019508

USP

Amikacin

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43N5O13
CAS Number:
Molecular Weight:
585.60
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

amikacin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N([C@H]1[C@@H]([C@H]([C@@H]([C@H](C1)N)O[C@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CN)O)O[C@H]2O[C@@H]([C@H]([C@@H]([C@H]2O)N)O)CO)C(=O)[C@@H](O)CCN

InChI

1S/C22H43N5O13/c23-2-1-8(29)20(36)27-7-3-6(25)18(39-22-16(34)15(33)13(31)9(4-24)37-22)17(35)19(7)40-21-14(32)11(26)12(30)10(5-28)38-21/h6-19,21-22,28-35H,1-5,23-26H2,(H,27,36)/t6-,7+,8-,9+,10+,11-,12+,13+,14+,15-,16+,17-,18+,19-,21+,22+/m0/s1

InChI key

LKCWBDHBTVXHDL-RMDFUYIESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amikacin USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Amikacin Sulfate
  • Amikacin Sulfate Injection
  • Kanamycin Injection
  • Kanamycin Sulfate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Bruce Montgomery et al.
Antimicrobial agents and chemotherapy, 58(7), 3714-3719 (2014-04-23)
The amikacin-fosfomycin inhalation system (AFIS) is a combination of 2 antibiotics and an in-line nebulizer delivery system that is being developed for adjunctive treatment of pneumonia caused by Gram-negative organisms in patients on mechanical ventilation. AFIS consists of a combination
Wenjuan Nie et al.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases, 25, 170-174 (2014-06-17)
To separate Mycobacterium abscessus subsp. bolletii from Mycobacterium abscessus subsp. abscessus using species identification, and to investigate the in vitro activity of amikacin, cefoxitin, imipenem, levofloxacin, moxifloxacin, clarithromycin, azithromycin, and linezolid against Mycobacterium abscessus. Seventy M. abscessus isolates, previously identified
Guilian Li et al.
PloS one, 10(2), e0119013-e0119013 (2015-02-20)
Isoniazid (INH) and rifampicin (RIF) are the two most effective drugs in tuberculosis therapy. Understanding the molecular mechanisms of resistance to these two drugs is essential to quickly diagnose multidrug-resistant (MDR) tuberculosis and extensive drug-resistant tuberculosis. Nine clinical Mycobacterium tuberculosis
Monika E Łysakowska et al.
Burns : journal of the International Society for Burn Injuries, 41(2), 364-371 (2014-08-26)
Acinetobacter baumannii isolates are responsible for a high number of wound infections. The reason of this study was to evaluate the activity of silver nanoparticles obtained by microexplosion against wide range of Acinetobacter spp. Susceptibility to silver nanoparticles was tested
Ming Zhang et al.
Antimicrobial agents and chemotherapy, 58(6), 3217-3223 (2014-03-26)
Oxazolidinones represent a new class of antituberculosis drugs that exert their function by inhibiting protein synthesis. Here, we compared the activities of three oxazolidinones, linezolid, PNU-100480, and AZD5847, against latent tuberculosis using a simple model employing the streptomycin-starved Mycobacterium tuberculosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service