Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3COOC(COOCH2CH3)(CH2COOCH2CH3)2
CAS Number:
Molecular Weight:
318.32
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1804947
grade
pharmaceutical primary standard
API family
triethyl citrate
manufacturer/tradename
USP
refractive index
n20/D 1.439 (lit.)
bp
228-229 °C/100 mmHg (lit.)
density
1.136 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
O=C(OCC)C(CC(OCC)=O)(CC(OCC)=O)OC(C)=O
InChI
1S/C14H22O8/c1-5-19-11(16)8-14(22-10(4)15,13(18)21-7-3)9-12(17)20-6-2/h5-9H2,1-4H3
InChI key
WEAPVABOECTMGR-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Acetyltriethyl citrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Storage Class
10 - Combustible liquids
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Friederike Eisenächer et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 88(3), 778-786 (2014-08-03)
Gastroretentive drug delivery systems are retained in the stomach for a sufficient time interval, releasing the drug in a controlled manner. According to literature, the floating principle is the most frequently used formulation approach for gastric retention. However, many publications
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1009923-500MG | 04061838665416 |
| 1009923-100MG | 04065268812006 |