Skip to Content
Merck
All Photos(1)

Documents

Z226211

Nucleosil® C8 HPLC Column

5 μm particle size, L × I.D. 15 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Agency

suitable for USP L7

Quality Level

product line

Nucleosil®

feature

endcapped: no

manufacturer/tradename

Nucleosil®

extent of labeling

8.5% carbon loading

parameter

400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

350 m2/g

matrix

octyl

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

100 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service