Key Documents
RAD125
radiello Anesthetic Gases and Vapor Sampler
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
manufacturer/tradename
Radiello
technique(s)
passive air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
This kit was developed for sampling of nitrous oxide, isofluorane, ethrane, halothane, and sevorane in surgical theaters. Parts for one complete sampler are packed separately in one sealed and sterile bag. Pack of 10 sterile bags.
Each sterile bag contains:
  • 1 permeative body
  • 1 support plate
  • 1 vertical adapter
  • 1 adsorbing cartridge
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
Rocco Rella et al.
The Science of the total environment, 487, 255-259 (2014-05-03)
The proofing treatment of the floor slabs with a solvent based material and successive treatment with epoxy resin resulted in significant air contamination in a primary school. The contamination caused illnesses in many occupants of school. The GC/MS investigations of
Anne Pennequin-Cardinal et al.
Talanta, 65(5), 1233-1240 (2008-10-31)
Radiello((R)) diffusive samplers filled with a thermally desorbable adsorbent (graphitised charcoal Carbograph 4) have been tested for the monitoring of BTEX. The sampling rates have been estimated under various controlled atmospheres in order to evaluate the effects of two factors
Articles
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Protocols
Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.
Related Content
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.