G005861
HPLC Analysis of Fat-Soluble Vitamins on Ascentis® Express C18, Methanol Mobile Phase
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase: methanol
flow rate: 1.5 mL/min
pressure: 3249 psi (224 bar)
column temp.: 30 °C
detector: UV, 280 nm
injection: 5 μL
sample: 0.5 mg/mL tocopherol acetate, alpha-tocopherol, delta-tocopherol and K1, 0.05 mg/mL D2, D3 and retinol acetate, 1.5 mg/mL retinol palmitate, all in ethanol.
suitability
application for HPLC
application(s)
vitamins, nutraceuticals, and natural products
Analysis Note
Legal Information
Analyte
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service