Skip to Content
Merck
All Photos(1)

Key Documents

CRM69174

Supelco

Acetochlor

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20ClNO2
CAS Number:
Molecular Weight:
269.77
Beilstein:
2859702
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

CCOCN(C(=O)CCl)c1c(C)cccc1CC

InChI

1S/C14H20ClNO2/c1-4-12-8-6-7-11(3)14(12)16(10-18-5-2)13(17)9-15/h6-8H,4-5,9-10H2,1-3H3

InChI key

VTNQPKFIQCLBDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Acetochlor belongs to the class of chloroacetamide herbicides, widely used against annual grass and broadleaf weeds. A long-chain fatty acid inhibitor, it is used for pre-emergence and pre-plant control in crops such as maize, peanuts, soybeans, cotton, potatoes, and sugarcane.
According to the Commission Implementing Regulation (EU) No 1372/2011 of 21st December 2011, acetochlor is not approved for use in the European Union (EU) as a plant protection agent, following Regulation (EC) No. 1107/2009. But maximum residue levels of 0.02 mg/kg and 0.05 mg/kg are allowed for its presence in frozen/fresh fruits and tree nuts respectively, according to the EU Reg. No 2015/603.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Acetochlor CRM may also be used as given below:


  • To develop a method for quantifying acetochlor and its metabolites in soybean and corn samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • QuEChERS-based sample preparation and ultra high-performance liquid chromatographic-tandem mass spectrometric (UHPLC-MS/MS) detection of clomazone and acetochlor residues in soybean samples
  • Analyze a modified QuEChERS-based sample extraction and clean-up method followed by validation using gas chromatography-tandem mass spectrometry (GC-MS/MS analysis) to determine 176 pesticide residues from different chemical classes in fatty matrices
  • Development of a sample treatment method based on ultrasound-assisted solvent extraction and solid-phase extraction clean-up for the quantification of five amide herbicides in fish samples by gas chromatography-electron capture detection (GC-ECD)
  • Simultaneous residue analysis of 42 pesticides in four egg samples using extraction based on the QuEChERS method and determination by UHPLC-MS/MS and GC-MS
  • Analysis of 20 herbicides in human blood samples by liquid-liquid extraction combined with ultra-pressure liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Kidney, Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of acetochlor on neurogenesis and behaviour in zebrafish at early developmental stages
Wang H, et al.
Chemosphere, 220, 954-964 (2019)
Determination of clomazone and acetochlor residues in soybean (Glycine max (L.) Merr.)
Hussan HNM, et al.
International Journal of Environmental Analytical Chemistry, 101(15), 2503-2509 (2021)
Simultaneous determination of 42 pesticides and herbicides in chicken eggs by UHPLC-MS/MS and GC-MS using a QuEChERS-based procedure
Li Y, et al
Chromatographia, 79(17), 1165-1175 (2016)
Development of an LC-MS/MS-based method for determination of acetochlor and its metabolites in crops
Saito-Shida, S, et al.
Journal of food composition and analysis, 108, 104454-104454 (2022)
Tao Huang et al.
Chemosphere, 243, 125345-125345 (2019-11-19)
Acetochlor is a high-volume herbicide used on a global scale and toxicity assessments are needed to define its potential for adverse effects in wildlife and humans. This study was conducted to determine the effects of acetochlor on human liver carcinoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service