Skip to Content
Merck
All Photos(1)

Key Documents

CRM46916

Supelco

Zearalenone solution

certified reference material, 50 μg/mL in acetonitrile, ampule of 1 mL

Synonym(s):

ZON

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O5
CAS Number:
Molecular Weight:
318.36
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

50 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

-10 to -25°C

SMILES string

OC1=C(C(O[C@@H](C)CCC2)=O)C(/C=C/CCCC2=O)=CC(O)=C1

InChI

1S/C18H22O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,20-21H,2,4-6,8-9H2,1H3/b7-3+/t12-/m0/s1

InChI key

MBMQEIFVQACCCH-QBODLPLBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.Zearalenone, also known as F-2 toxin, is produced by different Fusarium fungi, such as F. graminearum, F. culmorum, F. cerealis, F. equiseti, F. crookwellense, and F. semitectum. These fungi occur as regular contaminants in cereals such as corn, maize, and others. It is a secondary metabolite, known to have potent oestrogenic activity due to having structural similarity with endogenous estrogen.

Application

The CRM can also be used as follows:

  • Development of a fluorometric flow-through sensor for the determination of zearalenone in maize and maize-based animal feed samples
  • Multi-residue analysis of zearalenone and its five derivatives in edible vegetable oil samples by gas chromatography-triple quadrupole mass spectrometry (GC-QqQ-MS) following their gel permeation chromatography based extraction
  • Simultaneous determination of zearalenone and its derivatives from maize samples by magnetic solid-phase extraction (M-SPE) based on multi-walled carbon nanotube-magnetic nanoparticles (MWCNT-MNPs) and ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Construction of immunochromatographic test stripes based on colloidal gold sphere (AuSP) and nanorods (NRs) for the visual identification of zearalenone in cereal samples
  • Isotope dilution-liquid chromatography-tandem mass spectrometric (ID-LC-MS/MS) determination of zearalenone and its five metabolites in corn samples

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service