Skip to Content
Merck

A437

Bottom-of-the-Bottle filter/sparger

pore size 2 μm

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
SB.52
UNSPSC Code:
41115700
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

PEEK

packaging

pkg of 1 ea

manufacturer/tradename

Upchurch Scientific, Inc A-437

diam.

23 mm

pore size

2 μm

compatibility

configured for 1/8 in. O.D. tubing

Features and Benefits

Small enough to fit narrow neck bottles.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Bottom-of-the-Bottle is a trademark of IDEX Health & Science, LLC


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library