Skip to Content
Merck
All Photos(1)

Key Documents

59610

Supelco

SUPELCOSIL LC-ABZ (5 µm) HPLC Columns

L × I.D. 2 cm × 2.1 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm

Agency

suitable for USP L60

Quality Level

packaging

pkg of 1 kit

technique(s)

HPLC: suitable

L × I.D.

2 cm × 2.1 mm

matrix active group

amide, alkyl phase

particle size

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Footnote

Kit includes one cartride, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Korakas et al.
Journal of pharmaceutical and biomedical analysis, 12(9), 1141-1145 (1994-09-01)
A series of biologically active peptides and their conjugates with lipidic amino acids were investigated by systematic change of the mobile phase composition using traditional octadecylsilica stationary phase and the newly developed Supelcosil LC-ABZ column. The mobile phases contained various
J A López et al.
Journal of chromatography. A, 870(1-2), 105-114 (2000-03-18)
A high-performance liquid chromatographic method with electrochemical detection has been developed for the determination of six 1,4-dihydropyridines: nifedipine, nimodipine, nisoldipine, nicardipine, felodipine and lacidipine. The chromatographic separation was performed using a Supelcosil LC-ABZ+Plus C18 column. A mobile phase of methanol-water
Q C Li et al.
Journal of chromatographic science, 38(6), 246-254 (2000-07-13)
Nevirapine (Viramune), a dipyridiodiazepinone, is a potent and highly specific nonnucleoside inhibitor of HIV-1 reverse transcriptase. This paper describes the validation of a specific, sensitive, and stability-indicating high-performance liquid chromatography method for the assay and determination of related organic impurities
Murali Pendela et al.
Journal of separation science, 32(20), 3418-3424 (2009-09-19)
Streptomycin (SM) is composed of streptidine, streptose and N-methyl glucosamine sugar moieties. For the determination of SM and its related substances, an ion-pair LC-UV detection method using a Supelcosil LC-ABZ column was developed previously. While analyzing commercial samples, several unknown
Ana B Baranda et al.
Journal of chromatography. A, 1031(1-2), 275-280 (2004-04-03)
A high-performance liquid chromatographic method with electrochemical detection was developed for the simultaneous determination of five 1,4-dihydropyridines: amlodipine, nitrendipine, felodipine, lacidipine and lercanidipine. These drugs are widely used in the treatment of hypertension, angina pectoris and the therapy of cerebrovascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service