Skip to Content
Merck

58985

SUPELCOSIL LC-18 (3 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
NB.21
L × i.d.:
15 cm × 4.6 mm
Particle size:
3 μm
Matrix active group:
C18 (RP18), C18 (octadecyl) phase
Pore size:
120 Å
Matrix:
silica gel, spherical particle platform
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

SUPELCOSIL LC-18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm

agency

suitable for USP L1

Quality Segment

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

11.0% carbon loading

parameter

≤70 °C temp. limit, 400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.1 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

C18 (RP18), C18 (octadecyl) phase

particle size

3 μm

pore size

120 Å

pH

2-7.5

application(s)

food and beverages

compatibility

for use with DABTH-AA

separation technique

reversed phase

General description

A general purpose hydrophobic alkyl phase that is very retentive and gives good peak shape for a wide variety of compounds.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC


Still not finding the right product?

Explore all of our products under SUPELCOSIL LC-18 HPLC Column




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library