Skip to Content
Merck
All Photos(1)

Key Documents

58984C40

Supelco

SUPELCOSIL LC-18 (3 µm) HPLC Columns

L × I.D. 7.5 cm × 4 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

SUPELCOSIL LC-18 HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 4 mm

material

stainless steel column

Agency

suitable for USP L1

product line

SUPELCOSIL

feature

endcapped

manufacturer/tradename

SUPELCOSIL

packaging

1 ea of

extent of labeling

11.0% carbon loading

parameter

≤70 °C temp. limit
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 4 mm

surface area

170 m2/g

surface coverage

surface coverage 3.1 μmol/m2

matrix

silica gel, spherical particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

3 μm

pore size

120 Å

pH

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

A general purpose hydrophobic alkyl phase that is very retentive and gives good peak shape for a wide variety of compounds.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service