Skip to Content
Merck
All Photos(1)

Documents

581522-U

Supelco

Ascentis® Si (3 µm) HPLC Columns

L × I.D. 3 cm × 2.1 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Ascentis® Si HPLC Column, 3 μm particle size, L × I.D. 3 cm × 2.1 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L3

product line

Ascentis®

feature

endcapped: no

manufacturer/tradename

Ascentis®

packaging

1 ea of

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

3 cm × 2.1 mm

surface area

450 m2/g

impurities

<5 ppm metals

matrix

fully porous particle
silica gel high purity, spherical

matrix active group

silica phase

particle size

3 μm

pore size

100 Å

operating pH range

2-6

application(s)

food and beverages

separation technique

hydrophilic interaction (HILIC)
normal phase

Looking for similar products? Visit Product Comparison Guide

General description

The Ascentis family of columns is the fourth generation of HPLC column technology from Supelco scientists. Ascentis columns are bonded on high purity, 100 Angstrom silica including 3, 5, and 10 micron particle size. Columns are designed for small molecule applications and are scalable from micro columns (1.0 mm I.D.) to preparative dimensions (50 mm I.D.). The family includes C18, C8, Phenyl, Si and embedded polar group phase, RP-Amide.

The Ascentis Si is a high loading capacity silica with excellent peak shape. The Ascentis Si performs in both normal-phase and HILIC/ANP (aqueous normal phase) mode.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Szabolcs Fekete et al.
Journal of pharmaceutical and biomedical analysis, 49(1), 64-71 (2008-11-29)
The performance of 5 cm long columns packed with shell particles was compared to totally porous sub-2 microm particles in gradient and isocratic elution separations of hormones (dienogest, finasteride, gestodene, levonorgestrel, estradiol, ethinylestradiol, noretistherone acetate, bicalutamide and tibolone). Peak capacities
Szabolcs Fekete et al.
Journal of pharmaceutical and biomedical analysis, 50(5), 703-709 (2009-06-30)
The performance of a narrow bore silica based monolith column (5 cm x 2 mm) was compared to 5 cm long narrow bore (internal diameter < or = 2.1 mm) columns, packed with shell particles (2.7 microm) and totally porous
Shigehisa Uchiyama et al.
Analytical chemistry, 80(9), 3285-3290 (2008-03-18)
A new method for the simultaneous determination of ozone and carbonyls in air using a two-bed cartridge system has been developed. Each bed consists of reagent-impregnated silica particles. The first contains trans-1,2-bis-(4-pyridyl) ethylene (BPE) while the second contains 2,4-dinitrophenylhydrazine (DNPH).
Paweł Kubica et al.
Journal of chromatography. A, 1289, 13-18 (2013-04-04)
Two analytical procedures are proposed where HILIC and RPLC techniques are coupled with tandem mass spectrometry detection for rapid determination of trace amounts of nicotine in zero-level liquids for electronic cigarettes. Samples are prepared on the basis of the approach
Luana Magalhães et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1008, 98-107 (2015-12-08)
Nucleoside triphosphate diphosphohydrolase (NTPDase) is an enzyme belonging to the apyrase family that participates in the hydrolysis of the nucleosides di- and triphosphate to the corresponding nucleoside monophosphate. This enzyme underlies the virulence of parasites such as Leishmania. Recently, an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service