About This Item
UNSPSC Code:
41115700
NACRES:
NB.21
material
PEEK unit
packaging
pkg of 10
manufacturer/tradename
Upchurch Scientific, Inc F301X
I.D.
0.0625 in. (1.5875 mm)
fitting
1/4-28 nut and ferrule
General description
The one-piece fitting, 55067-U, is convenient to use because the ferrule will not stick in a receiving port and can be easliy found if dropped onto the floor. The two-piece fitting features a separate ferrule so you can replace just the ferrule instead of the entire unit. The Upchurch PEEK Fingertight fittings are for use with 1/16″ O.D. tubing and can be used to 6,000 psi (420 kg/cm2).
Still not finding the right product?
Explore all of our products under Upchurch PEEK Fingertight Fitting
Choose from one of the most recent versions:
Already Own This Product?