57217
Supelclean™ ENVI-Chrom P SPE Bulk Packing
pkg of 50 g
About This Item
Recommended Products
packaging
pkg of 50 g
technique(s)
solid phase extraction (SPE): suitable
surface area
900 m2/g
matrix
styrene/divinylbenzene
matrix active group
polystyrene-divinylbenzene phase
diameter
80-160 μm
pore size
20-300 Å
application(s)
food and beverages
separation technique
reversed phase
Related Categories
General description
Retention Mechanism: Reversed-phase or Adsorption
Sample Matrix Compatibility: Aqueous solutions
- Particle Size: 80-160 μm; Spherical Shape; Pore Size: 110- 175Å; Surface Area: 900m2/g
- Highly crosslinked, neutral, specially cleaned styrenedivinylbenzene resin used to retain hydrophobic compounds with some hydrophilic functionality under reversed phase conditions
- Highly resistant to extreme pH conditions
- Typical applications include aromatics and phenolic compounds from aqueous sample matrices
- Used for priority pollutant phenols from aqueous samples
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Reversed-phase interaction will retain most molecules with hydrophobic character; it is very useful for extracting analytes that are very diverse in structure within the same sample.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service