56102
Frit for Omnifit® Column Endpiece
polyethylene, for use with 10 mm Bore Columns, pore size 25 μm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
Recommended Products
material
polyethylene
packaging
pkg of 20 ea
manufacturer/tradename
Omnifit 6141
technique(s)
LPLC: suitable
pore size
25 μm
compatibility
for use with 10 mm Bore Columns
Looking for similar products? Visit Product Comparison Guide
General description
End pieces are equipped with porous frits. These can be made up of sintered metal, glass and plastic. The pore diameter and surface structures of the frits should be optimised such as to avoid clogging and to prevent abrasion of the chromatographic particles. The frits are made such that, they are easily exchangeable. Frits can also be used to keep the packing material in place.
Application
Polyethylene frits may be used with glass columns to conduct filtration experiments with bacterial suspensions. PTFE frits may also be used for preparative chromatography and solid phase extraction for borosilicate columns.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jan-Christer Janson
Protein Purification: Principles, High Resolution Methods, and Applications, 19-19 (2012)
Improvements to the pretreatment of bone at Oxford.
Ramsey, C. Bronk, et al.
Radiocarbon, 46, 155-164 (2004)
Population heterogeneity affects transport of bacteria through sand columns at low flow rates.
Simoni, Stefano F., et al.
Environmental Science & Technology, 32, 2100-2105 (1998)
L A Colón et al.
Journal of chromatography. A, 887(1-2), 43-53 (2000-08-29)
Considering the current interest in capillary electrochromatography (CEC), performed in packed columns, we present the different methods used to pack capillary columns for use in CEC. General considerations on column packing are given and the column fabrication process is discussed
Subperiosteal osteoid osteoma of the distal phalanx of the fourth toe.
M L LaCroix et al.
Orthopedics, 24(7), 695-696 (2001-08-02)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service