Skip to Content
Merck
All Photos(1)

Key Documents

48568

Supelco

Fluorene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10
CAS Number:
Molecular Weight:
166.22
Beilstein:
1363491
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

298 °C (lit.)

mp

111-114 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

C1c2ccccc2-c3ccccc13

InChI

1S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8H,9H2

InChI key

NIHNNTQXNPWCJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorene has been used as a test material in studying its electronic transitions in gas and crystalline phases, and in stretched polymer sheets, measured with synchrotron radiation.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

303.8 °F - closed cup

Flash Point(C)

151.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Electronic transitions of fluorene, dibenzofuran, carbazole, and dibenzothiophene: From the onset of absorption to the ionization threshold.
Nguyen D D, et al.
Journal of Molecular Spectroscopy, 264(1), 19-25 (2010)
C Grazia Bezzu et al.
Advanced materials (Deerfield Beach, Fla.), 24(44), 5930-5933 (2012-09-11)
A highly gas-permeable polymer with enhanced selectivities is prepared using spirobifluorene as the main structural unit. The greater rigidity of this polymer of intrinsic microporosity (PIM-SBF) facilitates gas permeability data that lie above the 2008 Robeson upper bound, which is
Rui Lu et al.
Environmental science and pollution research international, 20(4), 2220-2225 (2012-07-18)
Polycyclic aromatic hydrocarbons (PAHs) are persistent, bioaccumulative, and toxic chemicals and are listed as priority pollutants by the US EPA. Although they are sparsely soluble in water, their solubility can be increased by binding to dissolved organic matter in natural
Fu-Lin Sun et al.
Ecotoxicology (London, England), 21(6), 1651-1660 (2012-06-16)
Polycyclic aromatic hydrocarbons (PAHs) are of great environmental and human health concerns due to their widespread occurrence, persistence and carcinogenic properties. There is now compelling evidence that the mangrove sediment microbial structure is susceptible to PAHs contamination. The study aimed
Qianling Cui et al.
ACS applied materials & interfaces, 5(1), 213-219 (2012-12-15)
In the present work, a facile one-pot method is designed to fabricate a core-shell fluorescent nanoparticle (NP) for cellular imaging based on a new cationic conjugated polymer, poly[9,9'-bis(6,6'-(N,N,N-trimethylaminium)fluorene-2,7-ylenevinylene-co-alt-2,5-dicyano-1,4-phenylene] (PFVCN). Gold nanoflowers (AuNFs) are prepared by a seedless method, in which

Articles

Highly accurate HPLC-FLD method for evaluating 15 PAHs in olive oil using Ascentis® Express PAH HPLC column and SPE cleanup, following GB 5009.265-2021.

Highly accurate HPLC-FLD method for evaluating 15 PAHs in olive oil using Ascentis® Express PAH HPLC column and SPE cleanup, following GB 5009.265-2021.

Highly accurate HPLC-FLD method for evaluating 15 PAHs in olive oil using Ascentis® Express PAH HPLC column and SPE cleanup, following GB 5009.265-2021.

Highly accurate HPLC-FLD method for evaluating 15 PAHs in olive oil using Ascentis® Express PAH HPLC column and SPE cleanup, following GB 5009.265-2021.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service