Skip to Content
Merck
All Photos(1)

Key Documents

48418

Supelco

p-Terphenyl-d14 solution

certified reference material, 2000 μg/mL in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])-c2c([2H])c([2H])c(c([2H])c2[2H])-c3c([2H])c([2H])c([2H])c([2H])c3[2H]

InChI

1S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D

InChI key

XJKSTNDFUHDPQJ-WZAAGXFHSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ou-Ya Zhao et al.
Chemosphere, 174, 774-780 (2017-02-16)
The present study used strain ZH-H2 (Fusarium sp.) isolated by our group as the PAH-degrading strain and 5-6-rings PAHs as degradation objects. The soil incubation experiment was carried out to investigate the starch-enhanced degradation effects of HMW PAHs by Fusarium
Xiaoxue Zhang et al.
Environmental science and pollution research international, 27(34), 42969-42978 (2020-07-30)
The capacity of Fusarium sp. strain ZH-H2 to secret lignin peroxidase (LiP), laccase (Lac), and manganese peroxidase (MnP) and degrade high molecular weight polycyclic aromatic hydrocarbons (HMW-PAHs) was studied. When the fungus was grown in control mineral salt medium for

Articles

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service