Skip to Content
Merck
All Photos(1)

Key Documents

47783

Supelco

(±)-α-Tocopherol

analytical standard

Synonym(s):

DL-all-rac-α-Tocopherol, Vitamin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Beilstein:
94012
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E307

grade

analytical standard

Quality Level

form

liquid

CofA

current certificate can be downloaded

feature

standard type fat soluble vitamin

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.950 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

-10 to -25°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3

InChI key

GVJHHUAWPYXKBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Tocopherol is a form of fat-soluble vitamin E and shows anti-oxidant, anti-inflammatory, anti-cell proliferative, and anti-cancer properties.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

The analytical standard can also be used as follows:

  • Simultaneous determination of vitamin A and all four tocopherol isomers in human serum samples by ultra-high performance liquid chromatography (UHPLC) combined with fluorescence detection following their liquid-liquid extraction (LLE)
  • Multi-residue analysis of vitamin B (B1, B2, B5, and B9), and vitamin E (α-tocopherol, β-tocopherol, α-tocotrienol, and β-tocotrienol) in grass and grain samples of four different wheat varieties by liquid chromatography-electrospray ionization/multistage mass spectrometry (LC-ESI-MS/MS)
  • Separation and estimation of four tocopherols and four tocotrienols in pecan nut cultivar samples using non-aqueous reversed-phase high-performance liquid chromatography (NARP-HPLC) coupled with atmospheric pressure chemical ionization-tandem mass spectrometric (APCI-MS/MS) detection
  • Analysis of different cold pressed oil samples to detect and quantify eight tocol- homologs using a chemometrically-assisted optimized high-performance liquid chromatography (HPLC) with fluorescence detection (FLD) based method
  • Multi-residue determination of tocopherols in aromatic peanut oil (APO) and four refined vegetable oils using high-performance liquid chromatography with UV detection to detect adulteration

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of retinol, ?-tocopherol and ?-carotene in serum by isocratic high-performance liquid chromatography
Arnaud.J, et al.
Journal of Chromatography. B, Biomedical Applications, 572(1-2), 103-116 (1991)
Simultaneous determination of plasma retinol, alpha-tocopherol, lycopene, alpha-carotene, and beta-carotene by high-performance liquid chromatography.
Miller WK, et al.
Analytical Biochemistry, 138(2), 340-345 (1984)
Guowei Zheng et al.
Plant diversity, 42(1), 44-51 (2020-03-07)
Understanding the responses of crops to elevated atmospheric carbon dioxide concentrations (E[CO2]) is very important in terms of global food supplies. The present study investigates the effects of CO2 enrichment (to 800 μmol mol-1) on the physiology of soybean plants and the
George Gachumi et al.
Molecules (Basel, Switzerland), 26(5) (2021-04-04)
Phytosterols and tocopherols are commonly used in food and pharmaceutical industries for their health benefits. Current analysis methods rely on conventional liquid chromatography, using an analytical column, which can be tedious and time consuming. However, simple, and fast analytical methods
Kirsten G Coupland et al.
Movement disorders : official journal of the Movement Disorder Society, 29(13), 1606-1614 (2014-01-01)
Parkinson's disease (PD) is a neurodegenerative disorder for which environmental factors influence disease risk and may act via an epigenetic mechanism. The microtubule-associated protein tau (MAPT) is a susceptibility gene for idiopathic PD. Methylation levels were determined by pyrosequencing of

Protocols

HPLC Analysis of Fat Soluble Vitamins by Normal Phase Chromatography on Ascentis® S

Separation of δ-Tocopherol, analytical standard; Retinyl acetate; (+)-γ-Tocopherol; α-Tocopherol, ≥95.5%; Retinol; Vitamin A (acetate), meets USP testing specifications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service