Skip to Content
Merck
All Photos(1)

Key Documents

442794

Supelco

tert-Amyl methyl ether

analytical standard

Synonym(s):

1,1-Dimethylpropyl methyl ether, Methyl 2-methyl-2-butyl ether, Methyl tert-pentyl ether, TAME

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2OCH3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731452
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.389 (lit.)
n20/D 1.3896

bp

85-86 °C (lit.)

density

0.76-0.78 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

CCC(C)(C)OC

InChI

1S/C6H14O/c1-5-6(2,3)7-4/h5H2,1-4H3

InChI key

HVZJRWJGKQPSFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Amyl methyl ether (TAME) is a fuel oxygenate which is utilized as an additive in gasoline to enhance the octane rating and to improve combustion.[1][2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
TAME has been used as a reference standard for the determination of the analyte in Catalonian groundwater[3] and soil samples[4] by purge-and-trap gas chromatography/mass spectrometry (P&T-GC/MS) and in aqueous solutions by solid-phase microextraction (SPME) combined with GC-MS.[5] It may be used as a reference standard for the identification of the analyte in gasoline-contaminated groundwater by GC combined with various detectors/quantification methods such as photoionization detector (PID), flame ionization detector (FID), or MS.[6]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of standard methods for the analysis of methyl tert-butyl ether and related oxygenates in gasoline-contaminated groundwater
Halden RU, et al.
Environmental Science & Technology, 35(7), 1469 -11474 (2001)
Simultaneous determination of methyl tert-butyl ether, its degradation products and other gasoline additives in soil samples by closed-system purge-and-trap gas chromatography -mass spectrometry
Rosell M, et al.
Journal of Chromatography A, 1132(1-2), 28 -238 (2006)
Trace analysis of ethanol, MTBE, and related oxygenate compounds in water using solid-phase microextraction and gas chromatography/mass spectrometry
Cassada DA, et al.
Analytical Chemistry, 72(19), 4654 -44658 (2000)
Determination of low level methyl tert-butyl ether, ethyl tert-butyl ether and methyl tert-amyl ether in human urine by HS-SPME gas chromatography/mass spectrometry
Scibetta L, et al.
Analytica Chimica Acta, 581(1), 53 -562 (2007)
Single-walled carbon nanotubes as an effective adsorbent in solid-phase microextraction of low level methyl tert-butyl ether, ethyl tert-butyl ether and methyl tert-amyl ether from human urine
Rastkari N, et al.
Journal of Chromatography. B, Biomedical Applications, 877(14-15), 1568 -11574 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service