Skip to Content
Merck
All Photos(1)

Key Documents

442539

Supelco

1-Decanol

analytical standard

Synonym(s):

n-Decyl alcohol, Alcohol C10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9OH
CAS Number:
Molecular Weight:
158.28
Beilstein:
1735221
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

4.5 (vs air)

vapor pressure

1 mmHg ( 70 °C)
8.25 mmHg ( 100 °C)

CofA

current certificate can be downloaded

autoignition temp.

550 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.437 (lit.)

bp

231 °C (lit.)

mp

5-7 °C (lit.)

solubility

H2O: slightly soluble 0.0211 g/L at 20 °C

density

0.829 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCCCO

InChI

1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3

InChI key

MWKFXSUHUHTGQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Decanol can be obtained via decomposition of decyl hydroperoxide or dehydrogenation of 1-decene, followed by hydrolysis.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

1-Decanol can find applications as a solvent in order to recover morpholine, via extraction of morpholine from the aqueous effluent in rubber, petrochemical, dye, coking plants, resin and also organo-chemical industries.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - Pensky-Martens closed cup

Flash Point(C)

95 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Groundwater Chemicals Desk Reference (2010)
Isobaric phase equilibrium of morpholine+ 1-decanol, volumetric properties and molar refractivity from 293.15 to 333.15 K of morpholine+ 1-decanol and 1-octanol+ toluene system with applications of Prigogine?Flory?Patterson theory
Kumari A, et al.
Thermochimica Acta, 649, 41-53 (2017)
Shuo-Yang Wei et al.
Journal of chromatography. A, 1218(51), 9142-9148 (2011-11-22)
A novel method using sample preparation method, "ultrasound-assisted emulsification microextraction" (USAEME) with manual shaking, coupled with gas chromatography using and an electron capture detector (GC-ECD) was developed for the analysis of organochlorine pesticides (OCPs) in aqueous samples. The apparatus is
Grit Neumann et al.
Applied and environmental microbiology, 72(6), 4232-4238 (2006-06-06)
The solvent-tolerant strain Pseudomonas putida DOT-T1E was grown in batch fermentations in a 5-liter bioreactor in the presence and absence of 10% (vol/vol) of the organic solvent 1-decanol. The growth behavior and cellular energetics, such as the cellular ATP content
Christopher S Lunde et al.
Antimicrobial agents and chemotherapy, 53(8), 3375-3383 (2009-05-28)
Telavancin is an investigational lipoglycopeptide antibiotic currently being developed for the treatment of serious infections caused by gram-positive bacteria. The bactericidal action of telavancin results from a mechanism that combines the inhibition of cell wall synthesis and the disruption of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service