Skip to Content
Merck
All Photos(1)

Key Documents

40010-U

Supelco

1,1,1-Trichloroethane solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

Methyl chloroform

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)C

InChI

1S/C2H3Cl3/c1-2(3,4)5/h1H3

InChI key

UOCLXMDMGBRAIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Ozone 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K T Bogen et al.
Regulatory toxicology and pharmacology : RTP, 10(1), 26-50 (1989-08-01)
A methodology for using physiologically based pharmacokinetic (PBPK) models to derive predicted safe concentrations of noncarcinogens in drinking water for humans based on experimentally determined no observed adverse effect levels (NOAELs) in animals is presented and applied to the case
Lelio Zoccolillo et al.
Chemosphere, 76(11), 1525-1532 (2009-06-23)
For the purpose of understanding the transport and deposition mechanisms and the air-water distribution of some volatile chlorinated hydrocarbons (VCHCs), their atmosphere/aquatic environment concentration ratio was evaluated. In addition, for the purpose of differentiating VCHC behaviour in a temperate climate
Mathieu Valcke et al.
Inhalation toxicology, 23(14), 863-877 (2011-11-17)
The objective of this study was to assess the impact of the exposure duration and intensity on the human kinetic adjustment factor (HKAF). A physiologically based pharmacokinetic model was used to compute target dose metrics (i.e. maximum blood concentration (C(max))
Ariel Grostern et al.
Environmental microbiology, 12(4), 1053-1060 (2010-01-22)
Chloroform (CF), or trichloromethane, is an ubiquitous environmental pollutant because of its widespread industrial use, historically poor disposal and recalcitrance to biodegradation. Chloroform is a potent inhibitor of metabolism and no known organism uses it as a growth substrate. We
Keith L Shelton
The Journal of pharmacology and experimental therapeutics, 333(2), 612-620 (2010-02-04)
The present study examined the involvement of the GABAA, N-methyl-D-aspartate (NMDA), nicotinic acetylcholine, and mu-opioid receptor systems in the transduction of the discriminative stimulus effects of the abused inhalant 1,1,1-trichloroethane (TCE). Sixteen B6SJLF1/J mice were trained to discriminate 10 min

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service