Skip to Content
Merck

22005

SWAGELOK® Reducing Union

SWAGELOK® 400-6-2, brass, 1/4 in. SWAGELOK

Synonym(s):

Swagelok® 400-6-2,BR.REDUC.UNION 1/4X1/8 PK2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41115717
NACRES:
SB.54
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

brass

packaging

pkg of 2 ea

manufacturer/tradename

SWAGELOK® 400-6-2

fitting

1/4 in. SWAGELOK, 1/8 in. SWAGELOK

General description

Reducing unions of Swagelok® type are mostly used as metal column terminators. All original reducing unions have a bore in the centre of about 6mm in length and an internal diameter of 1.2mm. Moreover, reducing unions have a large bore at the non-reducing site. Modification is necessary in reducing union in order to minimize peak broadening, pressure resistance and to ease the installation process.
Nuts and ferrules included.

Application

Brass Swagelok® reducing union maybe used for providing connections for the glass tubing fitted in it to the stainless steel tubing by means of PTFE ferrules, in an elution technique used for the determination of solubility of gases like argon, helium, nitrogen and carbon dioxide in molten salts at pressures ranging between 0.5 to 2 atm.

Legal Information

SWAGELOK is a registered trademark of Swagelok Company


Still not finding the right product?

Explore all of our products under Swagelok® Reducing Union




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library