20508
Inlet Liner, for Packed GC (unpacked)
pkg of 5 ea
All Photos(1)
About This Item
UNSPSC Code:
41115719
Recommended Products
packaging
pkg of 5 ea
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
91.5 mm × 3.0 mm × 1.8 mm
General description
Deactivated (silanized) glass inlet liner for packed injection ports.
- Fits Agilent (5890, 6890, and 7890)
The liner is the second most important part of the inlet. It is the centrepiece of the inlet system, basically a glass tube between the septum and the column, where the sample is evaporated and brought into the gas phase. There are three most important parameters one should keep in mind while choosing a liner, firstly liner volume, considering a typical liner has a volume of 900μl, secondly liner treatment, one should always use a deactivated liner, so that the glass does not react with the analytes introduced in the liner, thirdly liner design, different designs are available like straight, tapered, baffled and without glass wool and one should be careful enough while choosing the design such that it is compatible with the desired injection mode.
Application
Inlet liner maybe used for the determination and evaluation of volatile components in food products. The vegetable oil maybe directly placed into the GC inlet liner packed with glass wool and then inserted into the heated inlet of the GC for analysis.
comparable product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Fire Debris Analysis.
Stauffer E
Science, 249-253 (2007)
Analysis of vegetable oils for flavor quality by direct gas chromatography.
Dupuy HP
Journal of the American Oil Chemists' Society, 54 (10), 445-449 (1977)
Fang-Zi Guo et al.
Environmental science and pollution research international, 23(20), 20506-20516 (2016-07-28)
Endosulfan is a persistent organic pollutant and widely used in agriculture as a pesticide. It is present in air, water, and soil worldwide; therefore, it is a health risk affecting especially the reproductive system. The aim of this study was