Skip to Content
Merck
All Photos(1)

Key Documents

15019AST

Supelco

Astec® CHIROBIOTIC® V2 Chiral (5 μm) HPLC Columns

L × I.D. 15 cm × 2.1 mm, HPLC Column

Synonym(s):

Chiral Stationary Phase Column V2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Astec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm

material

stainless steel column

Agency

suitable for USP L88

description

HPLC column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

15 cm × 2.1 mm

matrix

High-purity silica gel particle platform
fully porous particle

matrix active group

vancomycin phase

particle size

5 μm

pore size

200 Å

operating pH range

3.5-7.0

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

Neutral molecules, amides, acids, esters and amines show considerable enantioselectivity on these vancomycin-based CSPs. A wide variety of secondary and tertiary amines have been separated on the CHIROBIOTIC® V in the polar ionic mode. The CHIROBIOTIC® V has demonstrated many of the separation characteristics of protein-based stationary phases, but with exceptional stability and much higher sample capacity. Some chiral analytes have been resolved that have not been reported separated on any other chiral stationary phase. CHIROBIOTIC® V and V2 differ in their bonding chemistry the pore size of the support particle, giving them different selectivity and preparative capacity.

  • Bonded phase: Vancomycin
  • Operating pH range: 3.5 - 7.0
  • Particle diameter: 5, 10 or 16 μm
  • Pore size: 100 Å (CHIROBIOTIC® V) or 200 Å (CHIROBIOTIC® V2)

CHIROBIOTIC FAQs
CHIROBIOTIC Reference Bibliography
Chiral Product Literature

Application


  • Development and validation of an UFLC-MS/MS method for enantioselectivity determination of d,l-thero-methylphenidate, d,l-thero-ethylphenidate and d,l-thero-ritalinic acid in rat plasma and its application to pharmacokinetic study.: This study highlights the application of the Astec® CHIROBIOTIC® V2 chiral HPLC column in the pharmacokinetic assessment of chiral drugs, demonstrating its utility in accurate and efficient chiral separation for analytical chemistry in pharmacological research (Zhang et al., 2016).


  • New high-performance liquid chromatography method for the determination of (R)-warfarin and (S)-warfarin using chiral separation on a glycopeptide-based stationary phase.: This paper describes a method using the Astec® CHIROBIOTIC® V2 chiral HPLC column to differentiate between the enantiomers of warfarin, showcasing its critical role in therapeutic drug monitoring and clinical pharmacology, which is essential for ensuring patient safety and efficacy in treatments (Malakova et al., 2009).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatography Liquid Chiral Separations
Xiao, T.L., et al.
Encyclopedia of Separation Science, 1-15 (2000)
Validation of a chiral liquid chromatography?tandem mass spectrometry method for the determination of pantoprazole in dog plasma
Chen, Meixia, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 906, 85-90 (2012)
Doris Frühauf et al.
Journal of chromatography. A, 1269, 242-254 (2012-10-30)
A sub-minute enantioselective normal phase high performance liquid chromatographic (HPLC) method for the analysis of a chiral precursor molecule employed frequently in folic acid syntheses was developed, optimized and successfully validated according to ICH-guidelines. It could be shown that ultra-high
Enantioselective and sensitive determination of carvedilol in human plasma using chiral stationary-phase column and reverse-phase liquid chromatography with tandem mass spectrometry
Jiang, Juanjuan, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 960, 92-97 (2014)
New chemo-enzymatic approaches for the synthesis of (R)- and (S)-bufuralol
Nagy, Botond, et al.
Tetrahedron Asymmetry, 25 (18-19), 1316-1322 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service