Skip to Content
Merck
All Photos(1)

Key Documents

12019AST

Supelco

Astec® CHIROBIOTIC® T Chiral (5 μm) HPLC Columns

L × I.D. 15 cm × 2.1 mm, HPLC Column

Synonym(s):

TM=["Astec"] TM=["CHIROBIOTIC"] T Chiral Chromatography Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm

material

stainless steel column

Agency

suitable for USP L63

description

HPLC column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

15 cm × 2.1 mm

matrix

High-purity silica gel particle platform
fully porous particle

matrix active group

teicoplanin glycopeptide phase

particle size

5 μm

pore size

100 Å

operating pH range

3.8-6.8

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

CHIROBIOTIC® T and T2 have teicoplanin as the chiral selector. They offer unique selectivity for a number of classes of molecules, specifically underivatized α, β, γ and cyclic amino acids, N-derivatized amino acids, hydroxy-carboxylic acids, acidic compounds including carboxylic acids and phenols, small peptides, neutral aromatic analytes and cyclic aromatic and aliphatic amines. Separations normally obtained on a chiral crown ether or ligand exchange-type CSPs are also possible on the CHIROBIOTIC® T and T2, but in much simpler mobile phases, like water-alcohol. In addition, all of the known beta-blockers (amino alcohols), and dihydrocoumarins have been resolved. CHIROBIOTIC® T and T2 differ in their bonding chemistry and the pore size of the support particle, giving them different selectivity and preparative capacity.

  • Bonded phase: Teicoplanin
  • Operating pH range: 3.8 - 6.8
  • Particle diameter: 5, 10 or 16 μm
  • Pore size: 100 Å (CHIROBIOTIC® T) or 200 Å (CHIROBIOTIC® T2)

CHIROBIOTIC FAQs
CHIROBIOTIC Reference Bibliography
Chiral Product Literature

Application


  • Chiral liquid chromatography-mass spectrometry (LC-MS/MS) method development for the detection of salbutamol in urine samples: This research highlights the application of Astec® CHIROBIOTIC® T Chiral HPLC Column in sports medicine, specifically in the sensitive and accurate detection of salbutamol, a performance-enhancing beta-agonist, using advanced chiral chromatography techniques (Chan et al., 2016).

  • High-performance liquid chromatographic enantioseparation of 2-aminomono- and dihydroxycyclopentanecarboxylic and 2-aminodihydroxycyclohexanecarboxylic acids on macrocyclic glycopeptide-based phases: Demonstrating the utility of Astec® CHIROBIOTIC® T Chiral HPLC Column for enantiomeric purity assessment in pharmaceutical research, this study provides insights into the precise separation of complex amino acid derivatives critical for developing new therapeutic agents (Berkecz et al., 2009).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Abd el-Hady et al.
Journal of pharmaceutical and biomedical analysis, 37(5), 919-925 (2005-05-03)
A simple, sensitive and selective high performance liquid chromatographic method with UV detection for the chiral separation of racemic methotrexate (rac-Mtx) and enantiomeric purity of L-methotrexate in pharmaceutical formulations was developed and validated. The chiral separation was optimized studying both
Syntheses of racemic and non-racemic silicon- and germanium-containing a-amino acids of the formula type H2NCH(CH2ElR3)COOH (El=Si, Ge; R=organyl) and incorporation of d-H2NCH(CH2SiMe3)COOH and d-H2NCH(CH2GeMe3)COOH into biologically active decapeptides: a study on C/Si/Ge bioisosterism
Merget, Markus, et al.
Journal of Organometallic Chemistry, 628 (2), 183-194 (2001)
Berit P Jensen et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 865(1-2), 48-54 (2008-03-07)
A stereoselective liquid chromatography-tandem mass spectrometry assay was developed and validated for quantification of S- and R-metoprolol at concentrations of 0.5-50 microg/L in human plasma. Metoprolol was extracted from plasma by liquid-liquid extraction with ethyl acetate (82% recovery). Chromatographic separation
P Forssén et al.
Journal of chromatography. A, 1212(1-2), 89-97 (2008-10-31)
The shapes of elution profiles are often significantly influenced by the presence of strongly adsorbed additives in the mobile phase. This aspect needs to be considered in quantitative optimization of preparative chromatography. The theoretical study carried out here is based
Felikss Mutulis et al.
Bioorganic & medicinal chemistry, 15(17), 5787-5810 (2007-07-10)
Two hundred and ten tertiary amides were prepared on solid phase. Diamines were coupled to activated carboxylated Wang polymer, and the polymeric substituted benzyloxycarbonyl protected diamines obtained were reacted with aldehydes or ketones in trimethyl orthoformate giving resin attached Schiff

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service