Skip to Content
Merck
All Photos(1)

Documents

11049-U

Supelco

Tenax® Porous Polymer Adsorbent

matrix Tenax GR, 20-35 mesh, bottle of 500g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
SB.54

product name

Tenax® Porous Polymer Adsorbent, matrix Tenax GR, 20-35 mesh, bottle of 500 g

form

solid

packaging

bottle of 500 g

parameter

350 °C temp. limit

surface area

~25 m2/g

matrix

Tenax GR

particle size

20-35 mesh

density

~0.40 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

Tenax porous polymers, based on 2,6-diphenyl-p-phenylene oxide, is widely used as an adsorbent in both air collection and purge trap applications. Its unique structure provides alternate and desirable adsorption/desorption characteristics compared to other porous polymers.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents

Footnote

Tenax GR is a composite material containing 70% Tenax TA and 30% graphite carbon.

Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Retention characteristics of some volatile compounds on Tenax GR.
Dostal VKM and Churacek J.
Journal of Chromatography A, 642 (1), 379-382 (1993)
Handbook of Water Analysis.
Leo M, et al.
Technology & Engineering, 625-625 (2007)
Evaluation of Tenax-GR adsorbent for the passive sampling of volatile organic compounds at low concentrations.
Cao X and Hewitt N.
Atmos. Environ., Part A, 27 (12), 1865-1872 (1993)
Jae Hwan Lee et al.
Journal of the Air & Waste Management Association (1995), 56(11), 1503-1517 (2006-11-23)
Four popular thermally desorbable adsorbents used for air sampling (Tenax TA, Tenax GR, Carbopack B, and Carbopack X) are examined for the potential to form artifacts with ozone (O3) at environmental concentrations. The performance of these adsorbents for the ketone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service