Skip to Content
Merck

07313

Demeton-S-methyl-sulfon

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H15O5PS2
CAS Number:
Molecular Weight:
262.28
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Beilstein/REAXYS Number:
1712536
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

InChI

1S/C6H15O5PS2/c1-4-14(8,9)6-5-13-12(7,10-2)11-3/h4-6H2,1-3H3

InChI key

PZIRJMYRYORVIT-UHFFFAOYSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma aldrich.com

Demeton-S-methyl sulfone is an insecticide that belongs to the class of organophosphorus compounds, the major group of pest controllers used in agricultural production.

As per Regulation (EC) No 1107/2009, demeton-S-methyl-sulfon is not approved for use in European Union. Maximum residue levels (MRLs) have been set according to Regulation (EU) No 889/2012 for oxydemeton-methyl as a sum of oxydemeton-methyl and Demeton-S-methylsulfone expressed as oxydemeton-methyl for various products of plant and animal origin from 0.01 to 0.05 mg/kg.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Demeton-S-methyl sulfone CRM may also find its use as described below:
  • Validation of a method to monitor 11 organophosphorus residues in samples of banana, papaya, and bell pepper using QuEChERS extraction and high performance liquid chromatography with tandem mass spectrometry (HPLC-MS/MS)(1)
  • Simultaneous determination of four pesticides, aldicarb, demeton-S-methyl, fenamiphos, and methiocarb and their metabolites in grape juices using solid-phase extraction and liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Quantification and confirmation of 16 priority pesticides in baby foods and their comparison using HPLC-MS/MS and UPLC-MS/MS coupled with tandem quadrupole mass spectrometry
  • Development of reversed phase liquid chromatography-electrospray ionization-tandem mass spectrometry method for the determination of 52 pesticides in tobacco
  • Validation of two variations of the QuEChERS method for the determination of 120 pesticides and metabolites in cereal-based infant foods by LC-MS/MS with electrospray interface (ESI)

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Demeton-S-methyl-sulfon


pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Chronic 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library