ZRB2252
Anti-HLA-DPB1 Antibody, clone 1E16 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
HLA class II histocompatibility antigen DP(W4) beta chain, HLA class II histocompatibility antigen, DP beta 1 chain, MHC class II antigen DPB1
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1E16, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
purified by
using Protein A
species reactivity
human
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... HLA-DPB1(3115)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Human tonsil tissue lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HLA-DPB1 in Human tonsil tissue lysate.
Tested Applications
Flow Cytometry Analysis: 1 µg from a representative lot detected HLA-DPB1 in one million Human peripheral blood mononuclear cells (PBMC).
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HLA-DPB1 in HEK 293 cells.
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected HLA-DPB1 in human tonsil tissue sections.
Affinity Binding Assay: A representative lot of this antibody bound HLA-DPB1 peptide with a KD of 1.0 x 10-7 in an affinity binding assay.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Storage and Stability
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service