Skip to Content
Merck
All Photos(17)

Key Documents

ZRB2187

Sigma-Aldrich

Anti-LARP1 Antibody, clone 3K10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

La-related protein 1;La ribonucleoprotein domain family member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3K10, recombinant monoclonal

description

3K10 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 123.51 kDa
observed mol wt ~32 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... LARP1(23367)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3K10 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects La-related protein 1 (LARP1). It targets an epitope within 23 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal half of human La-related protein 1 (LARP1).

Application

Quality Control Testing

Evaluated by Western Blotting with His-ABP-tagged recombinant fragment of Human LARP1.

Western Blotting Analysis (WB): 1:10,000 dilution of this antibody detected His-ABP-tagged recombinant fragment of Human LARP1.

Tested Applications

Western Blotting Analysis: A representative lot of this antibody detected LARP1 in MOLT4 and HeLa cell lysates (1:10,000 dilution) and HeyA8, HeLa, OVCAR-8 wild type, and OVCAR-8 LARP1 KO cells (1:5000 dilution).

Immunocytochemistry Analysis: A representative lot of this antibody detected LARP1 in HepG2 cells (1:100 dilution), OVCAR-8 wild type and LARP1 knockdown cells (1:500 dilution) and OVCAR-8 wild type and LARP1 KO cells (1:2000 dilution).

Immunohistochemistry (Paraffin) Analysis: A representative lot detected LARP1 in human cerebellum tissue sections (1:1000 dilution) and human colorectal cancer tissue sections (1:100 and 1:200 dilutions).

Immunoprecipitation Analysis: A representative lot of this antibody was used to immunoprecipitate LARP1 from OVCAR-8 cancer cells.

Affinity Binding Assay:: A representative lot of this antibody bound LARP1 peptide with a KD of 2.3 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

La-related protein 1 (UniProt: Q6PKG0; also known as La ribonucleoprotein domain family member 1) is encoded by the LARP1 (also known as KIAA0731, LARP) gene (Gene ID: 23367) in human. La-related protein 1 (LARP1) is a cytoplasmic RNA-binding protein that promotes translation of specific classes of mRNAs downstream of the mTORC1 complex. It contains 1 HTH La-type RNA-binding domain (aa 397-487) and it associates with the mRNA 5′cap in an mTOR-dependent manner. It associates with mRNAs containing a 5′ terminal oligopyrimidine (5′TOP) motif, which is present in mRNAs encoding for ribosomal proteins and several components of the translation machinery. It is also shown to associate with actively translating ribosomes via interaction with polyadenylate-binding protein 1 (PABPC1/PABP) and stimulate translation of mRNAs containing a 5′TOP, thereby regulating cell growth and proliferation. LARP1 undergoes phosphorylation on multiple serine and threonine residues in response to active mTORC1. These phosphorylations are important for interaction with RPTOR and the mTORC1 complex. Phosphorylation also promotes dissociation from the 5′UTR of mRNA molecules with a 5′TOP motif. LARP1 levels are up-regulated in a number of hepatocellular carcinoma cell lines and liver cancer lesions, as well as in patients with hepatocellular carcinoma with a lower survival rate. Two isoforms of LARP1 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Fonseca, BD et al. (2015). J. Biol. Chem. 290(26):15996-6020; Tcherkezian, J., e tal. (2014). Genes Dev. 28(4); 357-371; Xie, C., t al. (2013). J. Transl. Med. 11; 272).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service