Z744111
Direct thermal Cryo-Tags® and Cryo-Spot combo
white, W × L 1.28 in. × 0.50 in. × 3/8 in., spot combo, pack of 1000 ea (500 tag and spot sets)
Synonym(s):
cryogenic vial labels, freezer labels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
white
packaging
pack of 1000 ea (500 tag and spot sets)
manufacturer/tradename
Diversified Biotech DTCR-6100
technique(s)
cryopreservation: suitable
W × L
1.28 in. × 0.50 in. × 3/8 in.
Looking for similar products? Visit Product Comparison Guide
General description
Temperature Range: 70°C to -196°C
Application
Cryogenics
Legal Information
Cryo-Tags is a registered trademark of Diversified Biotech
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service