Skip to Content
Merck
All Photos(1)

Key Documents

Z742614

G-CAPSULE Accessory Weight

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123100
NACRES:
NB.35
Pricing and availability is not currently available.

manufacturer/tradename

G-BioSciences 786-004

packaging

1 ea of

General description

G-CAPSULE Weight
  • Prevents G-CAPSULE from floating during electrophoresis
  • The G-CAPSULE Weight is designed to trap G-CAPSULE when submerged under buffer and prevents it from floating or changing direction during electroelution.  

Legal Information

G-Capsule is a trademark of Geno Technology, Inc.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tokinori Iwamoto et al.
The Journal of general virology, 86(Pt 10), 2807-2816 (2005-09-28)
Striped jack nervous necrosis virus (SJNNV), which infects fish, is the type species of the genus Betanodavirus. This virus has a bipartite genome of positive-strand RNAs, designated RNAs 1 and 2. A small RNA (ca. 0.4 kb) has been detected
K Tanaka et al.
Yeast (Chichester, England), 15(11), 1133-1139 (1999-08-24)
We cloned and sequenced homologs of RAS(CnRAS) and RHO1(CnRHO1) genes from Cryptococcus neoformans. The proteins encoded by the CnRAS and CnRHO1 genes contained 216 and 197 amino acids, respectively. The deduced amino acid sequence of the CnRAS gene shared a
E Dudley et al.
Microbiology (Reading, England), 147(Pt 1), 215-224 (2001-02-13)
Amino acid aminotransferases (ATases), which catalyse the last biosynthetic step of many amino acids, may have important physiological functions in Lactococcus lactis during growth in milk. In this study, the aspartate ATase gene (aspC) from L. lactis LM0230 was cloned
Sirshendu Chatterjee et al.
Acta biochimica et biophysica Sinica, 44(3), 259-268 (2012-01-20)
Sheep red blood cell (SRBC), a non-specific biological response modifier that has long been used as a classical antigen, has been shown to exert an immunomodulatory and anti-tumor activities in experimental animals. The active component of SRBC, which is responsible
S Braschi et al.
Biochimica et biophysica acta, 1392(2-3), 276-290 (1998-06-19)
Transgenic mice were created overproducing a range of human HL (hHL) activities (4-23-fold increase) to further examine the role of hepatic lipase (HL) in lipoprotein metabolism. A 5-fold increase in heparin releasable HL activity was accompanied by moderate (approx. 20%)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service