Z709204
ART® self-sealing barrier pipette tips
ART XLP 200, volume range 5-200 μL, sterile
Synonym(s):
pipet tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
maximum volume:
200 μL
volume range:
5-200 μL
sterility:
sterile
feature:
barrier
packaging:
pack of 3072 ea (4 packs of 768)
manufacturer/tradename:
Molecular BioProducts 2160P
Recommended Products
material
colorless
colorless tip
sterility
sterile
feature
barrier
packaging
pack of 3072 ea (4 packs of 768)
manufacturer/tradename
Molecular BioProducts 2160P
technique(s)
PCR: suitable
maximum volume
200 μL
volume range
5-200 μL
Looking for similar products? Visit Product Comparison Guide
General description
Extended length like the Art XLP but draws a full 200 μl
Application
ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).
Packaging
768EA = 8 trays of 96 tips (1 pack) and 3072EA = 4 Pack
960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
Compatibility
Compatibility
Biohit Proline-Variable 50-200 μl
Capp Aero 200 μl
Eppendorf Research/Reference 20-200 μl
Eppendorf Research Pro 5-100 μl
Eppendorf Response 4850 5-100 μl
Finnpipette 4027/4500 40-200 μl
Mettler-Toledo VoluMate 20-200 μl
Nichiryo BenchMate 20-200 μl
Pipetman P-200 20-200 μl
Rainin EDP 2 20-250 μl
Rainin EDP Plus 20-250 μl
Rainin Pipet Plus 20-200 μl
Rainin Pipetman 20-200 μl
Socorex/Wheaton 822 Series 10-100 μl
Biohit Proline-Variable 50-200 μl
Capp Aero 200 μl
Eppendorf Research/Reference 20-200 μl
Eppendorf Research Pro 5-100 μl
Eppendorf Response 4850 5-100 μl
Finnpipette 4027/4500 40-200 μl
Mettler-Toledo VoluMate 20-200 μl
Nichiryo BenchMate 20-200 μl
Pipetman P-200 20-200 μl
Rainin EDP 2 20-250 μl
Rainin EDP Plus 20-250 μl
Rainin Pipet Plus 20-200 μl
Rainin Pipetman 20-200 μl
Socorex/Wheaton 822 Series 10-100 μl
Other Notes
Browse all pipette tips
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service