Z709182
ART® self-sealing barrier pipette tips
ART 100, volume range 1-100 μL, sterile
Synonym(s):
pipet tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
maximum volume:
100 μL
volume range:
1-100 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
endotoxin free
RNase free
barrier
endotoxin free
packaging:
pack of 4800 ea (50 racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2065
Recommended Products
material
colorless tip
sterility
sterile
feature
DNase free
RNase free
barrier
endotoxin free
packaging
pack of 4800 ea (50 racks of 96 tips)
manufacturer/tradename
Molecular BioProducts 2065
technique(s)
PCR: suitable
maximum volume
100 μL
volume range
1-100 μL
Looking for similar products? Visit Product Comparison Guide
General description
Achieve precise and consistent results in your applications with ART® self-sealing barrier pipette tips in Lift-off Lid Rack. These tips are specially designed to effectively seal when exposed to potential contaminants, ensuring that they are trapped inside the barrier for enhanced sample protection. The ART barrier pipette tips are suitable for a wide range of pipetting applications.
Application
ART® self-sealing barrier pipette tips are ideal for loading gels, including acrylamide or agarose gels and for handling genomic DNA, fragile cell lines and other viscous materials.
ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).
Features and Benefits
- Certified sterile tips are free of RNase, DNase, and pyrogen contamination.
- Available in both sterile and nonsterile options, with a variety of packaging choices.
- Manufactured according to the highest industry quality standards.
- Extended length design prevents contamination by keeping the pipette shaft from entering the sample vessel.
- Solvent safe for acids, bases, and aggressive organic solvents.
Packaging
960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
Compatibility
Compatibility
Biohit mLINE 10-100 μl
Biohit Electronic Multi-Channel 5-100 μl
Biohit Multi-Channel 5-50 μl
Eppendorf Research Pro 4860 5-100 μl
Eppendorf Titerman 4908 30-300 μl
Finnpipette 4027/4500 Digital 5-40 μl
Finnpipette Multi-Channel 5-50 μl
Hamilton SoftGrip Precision 100 μl
Nichiryo BenchMate 5-50 μl
Nichiryo BenchMate Multi-Channel 5-50 μl
Socorex/Wheaton Calibra/822 10-100 μl
Biohit mLINE 10-100 μl
Biohit Electronic Multi-Channel 5-100 μl
Biohit Multi-Channel 5-50 μl
Eppendorf Research Pro 4860 5-100 μl
Eppendorf Titerman 4908 30-300 μl
Finnpipette 4027/4500 Digital 5-40 μl
Finnpipette Multi-Channel 5-50 μl
Hamilton SoftGrip Precision 100 μl
Nichiryo BenchMate 5-50 μl
Nichiryo BenchMate Multi-Channel 5-50 μl
Socorex/Wheaton Calibra/822 10-100 μl
Other Notes
Browse all pipette tips
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service