Skip to Content
Merck
All Photos(1)

Key Documents

Z660043

Bandelin TICKOPUR R 30 cleaning concentrate

neutral cleaner

Synonym(s):

neutral detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35
Pricing and availability is not currently available.

packaging

pkg of 1 L bottle
pkg of 2 L bottle
pkg of 5 L cannister

manufacturer/tradename

Bandelin 810

availability

available only in EU

General description

Neutral cleaner - gentle cleaning, anti-corrosive, neutral, pH 7, dosage 1 to 5%, 1 to 10 min.

Contamination:

Light drilling-, grinding-, polishing- and lapping residues, dust, soot, oily and greasy residues etc.

Objects to be cleaned:

Glass, ceramics, plastics, rubber, steel, stainless steel, non-ferrous-, precious- and light metals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service