Skip to Content
Merck
All Photos(1)

Key Documents

Z0292

Sigma-Aldrich

α-Zearalanol

~98% (HPLC), powder, mycotoxin

Synonym(s):

2,4-Dihydroxy-6-(6α,10-dihydroxyundecyl)benzoic acid μ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O5
CAS Number:
Molecular Weight:
322.40
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

α-Zearalanol, ~98% (HPLC)

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

~98% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

UHPLC: suitable

solubility

methanol: 1.96-2.04 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

C[C@H]1CCC[C@H](O)CCCCCc2cc(O)cc(O)c2C(=O)O1

InChI

1S/C18H26O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h10-12,14,19-21H,2-9H2,1H3/t12-,14+/m0/s1

InChI key

DWTTZBARDOXEAM-GXTWGEPZSA-N

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Mastri et al.
Journal of steroid biochemistry, 23(3), 279-289 (1985-09-01)
It is now well established that the mycotoxin zearalenone and some of its derivatives possess oestrogenic activity. In the present study, the binding characteristics of [3H]zearalanol (P-1496) to different classes of sites including [1] the oestrogen receptor, [2] the higher
Yosra Ayed et al.
Mutation research, 726(1), 42-46 (2011-09-06)
Zearalenone (ZEN) is a non-steroidal estrogenic mycotoxin produced by Fusarium fungi. It contaminates different components of the food chain and can cause serious economic and public health problems. The major metabolites of ZEN in various animal species are alpha- and
Zhanhui Wang et al.
Journal of molecular recognition : JMR, 24(5), 815-823 (2011-08-04)
The molecular recognition of hapten-antibody is a fundamental event in competitive immunoassay, which guarantees the sensitivity and specificity of immunoassay for the detection of haptens. The aim of this study is to investigate the correlation between binding ability of one
Michele Solfrizzo et al.
Analytical and bioanalytical chemistry, 401(9), 2831-2841 (2011-09-06)
Humans and animals can be simultaneously exposed through the diet to different mycotoxins, including aflatoxins, ochratoxin A, deoxynivalenol, zearalenone, and fumonisins, which are the most important. Evaluation of the frequency and levels of human and animal exposure to these mycotoxins
Rui Feng et al.
Biosensors & bioelectronics, 42, 367-372 (2012-12-06)
Nanoporous PtCo alloy was designed as an antibody carrier for preparation of a highly sensitive immunosensor. The immunosensor was constructed by assembling the capture zeranol antibody on thionine decorated graphene nanosheets modified glassy carbon electrode. With an enzyme-free immunosensor mode

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service