Skip to Content
Merck
All Photos(1)

Key Documents

YEAST1

Sigma-Aldrich

Yeast Transformation Kit

reagents for introducing plasmid DNA into yeast

Synonym(s):

lithium acetate yeast transformation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

grade

for molecular biology

Quality Level

usage

 kit sufficient for >100 standard transformations

technique(s)

transformation: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Sigma′s Yeast Transformation Kit contains all necessary reagents and controls for efficient transformation of yeast by the lithium acetate method.

Application

Suitable for transformation of any strain of yeast. Convenient, flexible and sensitive, positive transformants can be obtained with as little as 10 ng of DNA; the optimum efficiency is in the 0.1- 3 μg range.

Features and Benefits

  • Easy and ready-to-use
  • Requires as little as 10 ng of plasmid DNA
  • Flexibility for any strain of yeast
  • Sufficient for over 100 standard transformations

Components

The Yeast Transformation Kit contains:
  • Transformation Buffer; 100 ml; 100 mM lithium acetate, 10 mM Tris HCl, pH 7.6, and 1 mM EDTA
  • Plate Buffer; 100 ml; 40% PEG, 100 mM lithium acetate, 10 mM Tris HCl, pH 7.5, 1 mM EDTA
  • Deoxyribonucleic acid from salmon teste, 10 mg/ml; 2 x 1 ml
  • Control Yeast Plasmid DNA pRS316 carrying the ura gene; 10 μg
  • Yeast Synthetic Drop-out Medium Supplement Without Uracil; 1 g

Principle

Transformation with a plasmid complementing the mutated gene enables the transformant to grow on medium lacking the required component. Yeast cells are made competent for transformation by incubation in a buffered lithium acetate solution. Transformation is then carried out by incubating the cells together with transforming DNA and carrier DNA in a solution containing polyethylene glycol (PEG).

Kit Components Also Available Separately

Product No.
Description
SDS

  • D9156Deoxyribonucleic acid, single stranded from salmon testes, For hybridization 2 x 1SDS

  • Y1501Yeast Synthetic Drop-out Medium Supplements, without uracil 1 gSDS

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple and efficient procedure for transformation of yeasts.
R Elble
BioTechniques, 13(1), 18-20 (1992-07-01)
Miren Zumárraga et al.
Chemistry & biology, 14(9), 1052-1064 (2007-09-22)
Fungal laccases are remarkable green catalysts that have a broad substrate specificity and many potential applications in bioremediation, lignocellulose processing, organic synthesis, and more. However, most of these transformations must be carried out at high concentrations of organic cosolvents in
Widening the pH activity profile of a fungal laccase by directed evolution.
Torres-Salas P
Chembiochem, 14(8), 934-937 (2013)
Improved method for high efficiency transformation of intact yeast cells.
D Gietz et al.
Nucleic acids research, 20(6), 1425-1425 (1992-03-25)
Mechanistic and structural insights into the regioselectivity of an acyl-CoA fatty acid desaturase via directed molecular evolution.
Vanhercke T
The Journal of Biological Chemistry, 286(15), 12860-12869 (2011)

Articles

Transformation introduces exogenous DNA into cells, a fundamental genetic modification process demonstrated in Streptococcus pneumoniae.

Transformation introduces exogenous DNA into cells, a fundamental genetic modification process demonstrated in Streptococcus pneumoniae.

Transformation introduces exogenous DNA into cells, a fundamental genetic modification process demonstrated in Streptococcus pneumoniae.

Transformation introduces exogenous DNA into cells, a fundamental genetic modification process demonstrated in Streptococcus pneumoniae.

See All

Protocols

The selection of plasmids in yeast is based on the use of auxotrophic mutant strains, which cannot grow without a specific medium component (an amino acid, purine, or pyrimidine)

Yeasts are considered model systems for eukaryotic studies as they exhibit fast growth and have dispersed cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service