Skip to Content
Merck
All Photos(1)

Key Documents

X4753

Sigma-Aldrich

XCT790

≥98% (HPLC), solid

Synonym(s):

3-[4-(2,4-Bis-trifluoromethylbenzyloxy)-3-methoxyphenyl]-2-cyano-N-(5-trifluoromethyl-1,3,4-thiadiazol-2-yl)acrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H13F9N4O3S
CAS Number:
Molecular Weight:
596.42
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

yellow

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

COc1cc(ccc1OCc2ccc(cc2C(F)(F)F)C(F)(F)F)\C=C(/C#N)C(=O)Nc3nnc(s3)C(F)(F)F

InChI

1S/C23H13F9N4O3S/c1-38-17-7-11(6-13(9-33)18(37)34-20-36-35-19(40-20)23(30,31)32)2-5-16(17)39-10-12-3-4-14(21(24,25)26)8-15(12)22(27,28)29/h2-8H,10H2,1H3,(H,34,36,37)/b13-6+

InChI key

HQFNFOOGGLSBBT-AWNIVKPZSA-N

Application

XCT790 has been used:
  • as an estrogen-related receptor (ERR)α inverse agonist in C2C12 myotubes
  • as an estrogen-related receptor (ERR)α inverse agonist to elucidate decidualization functionality of ERRα in endometrial embryonic stem cells
  • as an autophagy inducer in human neuroblastoma SH-SY5Y and HeLa cell lines.

Biochem/physiol Actions

XCT 790 is a 5′adenosine monophosphate-activated protein kinase (AMPK) activator. It also acts as a proton ionophore and an uncoupler of oxidative phosphorylation in mitochondria. XCT790 impairs vascular endothelial growth factor (VEGF) and angiopoietin 2 (Ang-2) expression and exhibits suppression of endometrial tumor via estrogen-related receptor (ERRα) inhibition. XCT790 mediates cell cycle arrest and favors apoptosis in triple-negative breast cancer (TNBC).
XCT790 is a potent and specific inverse agonist of ERRα. XCT790 is selective; showing no significant antagonist activity on related nuclear receptors, such as ERRγ or ERα at concentrations below 10 μM. XCT790 inhibits the constitutive activity of ERRα in both biochemical and cell-based assays. The IC50 value is 300-500 nM in transient transfection assays using GAL4-ERR LBD or full-length ERR with the mSHP promoter.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PengMing Sun et al.
Cancer management and research, 10, 2521-2535 (2018-08-22)
To explore the targeted therapy of estrogen-related receptor α (ERRα) in endometrial cancer (EC) cells and its potential mechanisms. The mRNA and protein expression levels of ERRα and estrogen receptor α (ERα) were detected by qPCR and Western blotting in
Banu Eskiocak et al.
Biochemistry, 53(29), 4839-4846 (2014-07-08)
XCT 790 is widely used to inhibit estrogen-related receptor α (ERRα) activity as an inverse agonist. Here, we report that XCT 790 potently activates AMP kinase (AMPK) in a dose-dependent and ERRα-independent manner, with active concentrations more than 25-fold below
Sin-Aye Park et al.
British journal of cancer, 123(6), 988-999 (2020-06-24)
Gremlin-1 (GREM1), one of the bone morphogenetic protein antagonists, is involved in organogenesis, tissue differentiation and kidney development. However, the role of GREM1 in cancer progression and its underlying mechanisms remain poorly understood. The role of GREM1 in breast cancer
Zhenyu Xu et al.
Theranostics, 10(9), 4201-4216 (2020-04-01)
Enhanced intratumoral androgen biosynthesis and persistent androgen receptor (AR) signaling are key factors responsible for the relapse growth of castration-resistant prostate cancer (CRPC). Residual intraprostatic androgens can be produced by de novo synthesis of androgens from cholesterol or conversion from
G Vargas et al.
Oncogene, 38(7), 950-964 (2018-11-28)
Bone is the most common metastatic site for breast cancer. Estrogen-related-receptor alpha (ERRα) has been implicated in cancer cell invasiveness. Here, we established that ERRα promotes spontaneous metastatic dissemination of breast cancer cells from primary mammary tumors to the skeleton.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service