Skip to Content
WPB01421
Whirl-Pak® Blender Bag
capacity 1627  mL (55 oz), bag W × L × thickness 19 cm × 30 cm × 0.076 mm
Synonym(s):
Nasco B01421, homogenizer sample bags, sample bag, sample receptacle, sample storage bag
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.74
UNSPSC Code:
24111500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
clear linear low-density polyethylene (LLDPE)
sterility
sterile
packaging
pkg of 2000 ea (2 boxes of 1000EA)
manufacturer/tradename
Whirl-Pak® B01421
bag W × L × thickness
19 cm × 30 cm × 0.076 mm
capacity
1627  mL (55 oz)
General description
These bags are ideal for use in a homogenizer in the lab with all solid and liquid contents including but not limited to: food, protein, powder, pharmaceuticals, dairy, environmental and many more. Whirl-Pak® sterilized sampling bags are ideal for food & beverage, dairy, water, medical, veterinary, environmental, soil, and industrial markets as well as surface, forensic science, genetics, biomedical, and pharmaceutical sampling. Whirl-Pak® bags have been the professionals’ trusted sampling bag for over 60 years and offer puncture-proof closure tabs, guaranteed sterility, easy identification, and a leak-proof closure you can rely on.
Features and Benefits
  • Perforated for easy opening and to protect sterility
  • Designed for quick testing and disposal when the sample does not have to be retained
  • Bag can be frozen to any temperature, even with liquid nitrogen, but should not be used at temperatures above 180°F (82°C)
  • FDA and USDA Approved
  • Post-manufacturing sterilization
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Still not finding the right product?
Explore all of our products under Whirl-Pak® Blender Bag
Choose from one of the most recent versions:
Certificates of Analysis (COA)