Skip to Content
Merck
All Photos(7)

Key Documents

WH0006667M2

Sigma-Aldrich

Monoclonal Anti-SP1 antibody produced in mouse

clone 1A5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Sp1 transcription factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A5, monoclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SP1(6667)

General description

Sp1 protein was the first transcription factor to be cloned and characterized. It was first detected in HeLa cells on the basis of its ability to activate the SV40 early promoter transcription. Analysis of structure and function has revealed that Sp1 can be separated into discrete functional domains. The DNA-binding domain consists of three zinc fingers that specifically bind to the GC-box element.

Immunogen

SP1 (NP_612482, 89 a.a. ~ 198 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GTGELDLTATQLSQGANGWQIISSSSGATPTSKEQSGSSTNGSNGSESSKNRTVSGGQYVVAAAPNLQNQQVLTGLPGVMPNIQYQVIPQFQTVDGQQLQFAATGAQVQQ

Biochem/physiol Actions

Sp1 protein was shown to recognize and bind selectively to a GC-rich consensus sequence (GC-box: GGGCGG or CACCC) that presents in the promoter of several important cellular genes, including Simian vacuolating virus 40 (SV40) early, human immunodeficiency virus-1 (HIV-1), platelet-derived growth factor subunit B (PDGF-B), Myc, c-Src etc. In addition to transcription, Sp1 function has been linked to cell growth, cancer and Huntington disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie-Therese El-Daher et al.
Cell discovery, 4, 61-61 (2018-11-21)
A loss-of-function mutation in tetratricopeptide repeat domain 7A (TTC7A) is a recently identified cause of human intestinal and immune disorders. However, clues to related underlying molecular dysfunctions remain elusive. It is now shown based on the study of TTC7A-deficient and
Structures of zinc finger domains from transcription factor Sp1. Insights into sequence-specific protein-DNA recognition.
Narayan VA
The Journal of Biological Chemistry, 272(12), 7801-7809 (1997)
Decreased association of the transcription factor Sp1 with genes downregulated in Huntington's disease.
Chen-Plotkin AS
Neurobiology of Disease, 22(2), 233-241 (2006)
Mithramycin is a gene-selective Sp1 inhibitor that identifies a biological intersection between cancer and neurodegeneration.
Sleiman SF
The Journal of Neuroscience, 31(18), 6858-6870 (2011)
Qingyun Peng et al.
International journal of biological sciences, 16(15), 2974-2988 (2020-10-17)
Sepsis-induced myocardial dysfunction (SIMD) is a life-threatening complication caused by inflammation, but how it is initiated is still unclear. Several studies have shown that extracellular high mobility group box 1 (HMGB1), an important cytokine triggering inflammation, is overexpressed during the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service