Skip to Content
Merck
All Photos(2)

Key Documents

WGA2

Sigma-Aldrich

GenomePlex® Complete Whole Genome Amplification (WGA) Kit

Optimized kit with enzyme for amplifying a variety of DNA including FFPE tissue

Synonym(s):

Whole genome amplification kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NA.55

Quality Level

technique(s)

whole genome amplification: suitable

shipped in

wet ice

storage temp.

−20°C

General description

GenomePlex® Complete Whole Genome Amplification Kit utilizes a proprietary technology based on random fragmentation of genomic DNA and conversion of the resulting small fragments to PCR-amplifiable library molecules flanked by universal priming sites. WGA is achieved by PCR amplification of the library molecules using universal oligonucleotide primers. It contains everything required for whole genome amplification including an optimized enzyme, WGA DNA Polymerase. It can be used in several applications and is suitable for use with purified genomic DNA from a variety of sources including blood cards, whole blood, buccal swabs, soil, plant, and serum. It allows the researcher to generate a representative, ~500-fold amplification of genomic DNA.

Application

GenomePlex® Complete Whole Genome Amplification (WGA) Kit has been used in the amplification of DNA. This kit is also suitable for use with downstream applications including:
  • microarray analysis
  • SNP analysis
  • STR analysis
  • DNA archiving

Features and Benefits

  • Higher yield from minimal template: Amplification of nanogram amounts of genomic DNA to microgram yields (around 10 μg) in less than about three hoursrs
  • Nanograms of samples can be preserved at –20 °C for future use
  • Choose from a variety of DNA sources: whole blood, buccal swab, blood card, plant, soil, & formalin-fixed paraffin-embedded tissue (FFPE)
  • Whole-genome amplification (WGA) DNA polymerase increases the amplification accuracy
  • Whole-genome representation with no detectable allele bias
  • Compatible with many downstream applications such as TaqMan® assays, single nucleotide polymorphism (SNP) analysis, comparative genomic hybridization (CGH) analysis

Other Notes

The sequences of the universal primers provided in this kit are considered proprietary.

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
GenomePlex is a registered trademark of Takara Bio USA, Inc.
TaqMan is a registered trademark of Roche Molecular Systems, Inc.

Kit Components Also Available Separately

Product No.
Description
SDS

  • W4502Water, Nuclease-Free Water, for Molecular BiologySDS

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Chen et al.
Toxicology, 292(2-3), 63-70 (2011-11-15)
Furan, a widely used industrial compound, has been found in a number of heated food items. Furan is carcinogenic to rats and mice, but the mechanism behind its carcinogenic effect is still not well understood. In this study, we tested
M Korabecna et al.
Neoplasma, 63(3), 402-410 (2016-03-02)
Tubulocystic renal cell carcinoma (TRCC) represents a rare tumor with incidence lower than 1 % of all renal carcinomas. This study was undertaken to contribute to characterization of molecular signatures associated with TRCC and to compare them with the features of papillary
Levi Yant et al.
The Plant cell, 22(7), 2156-2170 (2010-08-03)
The Arabidopsis thaliana transcription factor APETALA2 (AP2) has numerous functions, including roles in seed development, stem cell maintenance, and specification of floral organ identity. To understand the relationship between these different roles, we mapped direct targets of AP2 on a
Comparative metagenomics of viral assemblages inhabiting four phyla of marine invertebrates
Gudenkauf BM and Hewson I
Frontiers in Marine Science, 3, 23-23 (2016)
Novel cell enrichment technique for robust genetic analysis of archival classical Hodgkin lymphoma tissues
Juskevicius D, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 98, 1487-1499 (2018)

Articles

aCGH technology evolves for high-res chromosomal analysis, needing a large DNA input per sample, advancing genetic research.

aCGH technology evolves for high-res chromosomal analysis, needing a large DNA input per sample, advancing genetic research.

aCGH technology enhances chromosomal analysis, requiring significant DNA input per sample.

aCGH technology enhances chromosomal analysis, requiring significant DNA input per sample.

See All

Protocols

GenomePlex® amplification improves DNA yield from soil samples, ensuring downstream analysis reliability despite potential nuclease damage.

GenomePlex® amplification improves DNA yield from soil samples, ensuring downstream analysis reliability despite potential nuclease damage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service