Skip to Content
Merck
All Photos(1)

Key Documents

W3144

Sigma-Aldrich

Wortmannin

from Penicillium funiculosum, ≥95% (HPLC), solution, PI3-kinase inhibitor

Synonym(s):

Antibiotic SL-2052

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24O8
CAS Number:
Molecular Weight:
428.43
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Wortmannin, Ready Made Solution, from Penicillium funiculosum, ≥95% (HPLC)

biological source

Penicillium funiculosum

Quality Level

Assay

≥95% (HPLC)

storage condition

protect from light

concentration

10 mM in DMSO (0.2 μm filtered)

shipped in

wet ice

storage temp.

−20°C

SMILES string

[o]1c2c3c(c1)C(=O)O[C@@H]([C@@]3(C4=C([C@H]5[C@](C[C@H]4OC(=O)C)(C(=O)CC5)C)C2=O)C)COC

InChI

1S/C23H24O8/c1-10(24)30-13-7-22(2)12(5-6-14(22)25)16-18(13)23(3)15(9-28-4)31-21(27)11-8-29-20(17(11)23)19(16)26/h8,12-13,15H,5-7,9H2,1-4H3/t12-,13+,15+,22-,23-/m0/s1

InChI key

QDLHCMPXEPAAMD-QAIWCSMKSA-N

Gene Information

Application

Wortmannin, Ready Made Solution has been used:
  • as a phosphoinositide 3-kinase (PI3K) pathway inhibitor
  • as an endoderm-inducing agent in stem cell selection and differentiation
  • as a autophagy inhibitor

Wortmannin, a low molecular weight membrane permeable hydrophobic fungal metabolite, is widely used to study and verify the involvement of phosphoinosidite 3-kinase (PI3K)/Akt in cell signaling cascades that regulate key processes such as cell survival and apoptosis. Wortmannin may also be used to block and study the functional roles of polo-like kinases involved in the regulation of mitosis.

Biochem/physiol Actions

Wortmannin is a hydrophobic fungal metabolite with a sterol-like structure. Inhibition of PI3K/Akt signal transduction cascade by wortmannin enhances apoptotic effects.
Wortmannin is a low molecular weight; hydrophobic fungal metabolite with a sterol-like structure produced by Penicilium fumiculosum. Inhibition of PI3K/Akt signal transduction cascade by wortmannin, enhances the apoptotic effects of radiation or serum withdrawal and blocks the antiapoptotic effect of cytokines. PI3K inhibition by wortmannin also blocks many of the short-term metabolic effects induced by insulin receptor activation. Research has demonstrated that wortmannin inhibits two enzymes from mitotical division, key regulators Polo-like kinase (Plk) family, Plk1 and Plk3.
Wortmannin is a low-molecular-weight hydrophobic fungal metabolite with a sterol-like structure produced by Penicillium funiculosum. It is a selective inhibitor of phosphoinosidite 3-kinase. It interferes with Akt signal transduction cascade, enhancing the apoptotic effects of radiation or serum withdrawal and blocks the antiapoptotic effect of cytokines. Wortmannin is membrane-permeable, thus facilitating whole-cell studies of G-protein-coupled receptor and receptor tyrosine kinase signalling pathways. PI3K inhibition by wortmannin also blocks many of the short-term metabolic effects induced by insulin receptor activation. Research has demonstrated that wortmannin inhibits two mitotic enzymes, key regulators in the Polo-like kinase (Plk) family, Plk1 and Plk3.
Wortmannin possesses anti-inflammatory properties. This mold metabolite is capable of preventing an intermediate step in transcytosis. In fisher rat thyroid (FRT) cells, wortmannin helps to block bidirectional transcytosis of ricin.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Physical form

Wortmannin Ready Made Solution supplied as a 10 mM 0.2 μm-filtered solution in DMSO.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibiting the RAS-PI3K Pathway in Cancer Therapy
The Enzymes, 34, 107-136 (2013)
Wortmannin, an inhibitor of phosphoinositide 3-kinase, inhibits transcytosis in polarized epithelial cells
Hansen SH, et al.
The Journal of biological chemistry, 270(47), 28425-28432 (1995)
Inhibition of autophagy blocks cathepsins-tBid-mitochondrial apoptotic signaling pathway via stabilization of lysosomal membrane in ischemic astrocytes
Zhou XY, et al.
Cell Death & Disease, 8(2), e2618-e2618 (2017)
Rac1 targeting suppresses p53 deficiency-mediated lymphomagenesis
Bosco EE, et al.
Blood, 115(16), 3320-3328 (2010)
Generation and selection of pluripotent stem cells for robust differentiation to insulin-secreting cells capable of reversing diabetes in rodents
Southard SM, et al.
Testing, 13(9), e0203126-e0203126 (2018)

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service