Skip to Content
Merck

Skip To

V7127

D-Val-Leu-Lys p-nitroanilide dihydrochloride

plasmin substrate, chromogenic, ≥98% (HPLC), powder

Synonym(s):

D-Val-Leu-Lys p-nitroanilide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C23H38N6O5 · 2HCl
Molecular Weight:
551.51
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

D-Val-Leu-Lys p-nitroanilide dihydrochloride, plasmin substrate

Quality Segment

assay

≥98% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, light yellow

storage temp.

−20°C

SMILES string

Cl.[N+](=O)([O-])c1ccc(cc1)NC(=O)C(NC(=O)C(NC(=O)C(N)C(C)C)CC(C)C)CCCCN

InChI

1S/C23H38N6O5.ClH/c1-14(2)13-19(28-23(32)20(25)15(3)4)22(31)27-18(7-5-6-12-24)21(30)26-16-8-10-17(11-9-16)29(33)34;/h8-11,14-15,18-20H,5-7,12-13,24-25H2,1-4H3,(H,26,30)(H,27,31)(H,28,32);1H

InChI key

ITWYYDDNWFVYSU-UHFFFAOYSA-N

Application

D-Val-Leu-Lys p-nitroanilide dihydrochloride has been used:
  • as substrate to measure the activity of plasminogen activator by a chromogenic assay[1]
  • as a p-nitroanilide (pNA) substrate to measure the specific activities of serine protease isoforms in Gloydius intermedius venom[2]
  • as a substrate in total plasmin assay using milk/cream samples[3]


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V0882M4765G6133
assay

≥98% (HPLC)

assay

≥98% (TLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

solubility

water: 50 mg/mL, clear, light yellow

solubility

water: 50 mg/mL, clear, colorless to yellow

solubility

DMF: soluble 25 mg/mL, clear, colorless to light yellow

solubility

water: 5 mg/mL, clear, yellow

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

300

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters