Skip to Content
Merck

V4507

Nalgene® cryogenic vials

capacity 1.2 mL, non-sterile, External threads

Synonym(s):

cryostorage vial, cryovial cryotube, Nalgene 5011-0012, cold storage sample tube, cold storage tube, cyro storage tube

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41121704
Capacity:
1.2 mL
Material:
colorless tube, conical bottom, high-density polyethylene closure, polypropylene , polypropylene tube
Sterility:
non-sterile
Packaging:
pack of 1000 ea
Feature:
External threads
Parameter:
-150 °C min. temp.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

colorless tube, conical bottom, high-density polyethylene closure, polypropylene , polypropylene tube

sterility

non-sterile

feature

External threads

packaging

pack of 1000 ea

manufacturer/tradename

Nalgene® 5011-0012

parameter

-150 °C min. temp.

technique(s)

cryopreservation: suitable

capacity

1.2 mL

tube diam.

12.7 mm

General description

Vials designed for exceptional ease of use and sample integrity in ultra-low temperature storage. Closure seals without gasket or O-ring, minimizing risk of contamination. The deep skirt of the closure allows single-handed aseptic technique without exposing fingers to vial contents. Ribs around the bottom of the vial interlock with wells in the Cryovial Racks so the vial will not turn when opening or closing. Bottoms of vials are skirted; vial stands without holder.

Sterile vials and closures are pre-assembled and radiation-sterilized. A white marking area, fill line and graduations are printed on the side. Certified sterile, non-cytotoxic and non-pyrogenic.

Note: Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex tubing. Such use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build-up resulting in possible explosion or biohazard release.

Packaging

Non-sterile vials and closures are packed separately. Vials have no printing or graduations.

Legal Information

CryoFlex is a trademark of Thermo Fisher Scientific or its subsidiaries
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nalgene® cryogenic vials




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library