Skip to Content
Merck

V2764

Anti-Vanilloid Receptor-1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-TRPV1, Anti-VR-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ARR, WB
Citations:
7
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 90 kDa

species reactivity

rat

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

microarray: suitable, western blot: 1:1,000 using whole cell extract of 293T cells expressing rat VR-1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPV1(7442)

General description

Vanilloid Receptor-1 (VR-1) is a cation channel protein that stimulates a burning pain sensation upon binding to capsaicin, a component of ′hot′ chilli peppers. VR-1 is also activated by noxious heat stimuli. Moreover, VR-1 can combine both, heat and chemical stimuli to cause a sensation of pain in vivo.

Immunogen

synthetic peptide corresponding to the C-terminus of rat vanilloid receptor-1 (amino acids 817-838) conjugated to KLH. The sequence is highly conserved in guinea pig (~85% identity) and human (~70% identity) and is not found in rat VRL-1.

Biochem/physiol Actions

Anti-Vanilloid Receptor-1 antibody is specific for vanilloid receptor-1 (90 kDa) in rats. The vanilloid receptor-1 immunizing peptide (rat, amino acids 817-838) specifically inhibits the staining of the vanilloid receptor-1 band in immunoblotting.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library