Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.79
UNSPSC Code:
41116107
Pricing and availability is not currently available.
assay
~98% (Components, TLC)
Quality Segment
form
powder
color
light yellow to dark yellow, faint yellow-brown to yellow-brown
storage temp.
−20°C
General description
Vitamins Kit is a multivitamin mix comprising biotin, folic acid, vitamin B6, riboflavin, thiamine, D-pantothenic acid and niacinamide.
Application
Vitamins Kit has been used as a vitamin supplement in the minimal medium for conidia spores and vegetative cultures.
Biochem/physiol Actions
Vitamins serve as a cofactor for enzymes and regulates many biological processes. They are essential component of cell culture medium. Vitamins A, E, B9 and B12 support immune system and have anti-inflammatory functionality. Vitamin B complex aids in nervous system improvement and coordinates energy production. Vitamin B1 is a micronutrient needed for cellular metabolic processes.
Other Notes
11 Vitamins in quantities as indicated:
A9878 p-Aminobenzoic Acid, 5 g
B4501 d-Biotin, 100 mg
F7876 Folic Acid, 1g
N3376 Niacinamide, 100 g
21210 D-Pantothenic Acid, hemicalcium saltt, 5 g
P9130 Pyridoxal Hydrochloride, 500 mg
P9380 Pyridoxamine Dihydrochloride, 250 mg
P9755 Pyridoxine Hydrochloride, 5 g
R4500 Riboflavin, 5 g
T4625 Thiamine Hydrochloride, 5 g
T5625 DL-6,8-Thioctic Acid, 500 mg
A9878 p-Aminobenzoic Acid, 5 g
B4501 d-Biotin, 100 mg
F7876 Folic Acid, 1g
N3376 Niacinamide, 100 g
21210 D-Pantothenic Acid, hemicalcium saltt, 5 g
P9130 Pyridoxal Hydrochloride, 500 mg
P9380 Pyridoxamine Dihydrochloride, 250 mg
P9755 Pyridoxine Hydrochloride, 5 g
R4500 Riboflavin, 5 g
T4625 Thiamine Hydrochloride, 5 g
T5625 DL-6,8-Thioctic Acid, 500 mg
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form powder | form solution | form liquid | form solution |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| assay ~98% (Components, TLC) | assay 98% (CP) | assay 97% (CP Sum of E & Z Isomers) | assay 98% (CP) |
| color light yellow to dark yellow, faint yellow-brown to yellow-brown | color colorless | color yellow to orange | color - |
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






