Skip to Content
Merck
All Photos(1)

Key Documents

UC448

Sigma-Aldrich

Paracetamol sulfate potassium salt

solid, ≥97% (HPLC)

Synonym(s):

4-Acetamidophenol sulfate ester potassium salt, Acetaminophen sulfate potassium salt, N-(4-Sulfoxyphenyl)acetamide monopotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8KNO5S
CAS Number:
Molecular Weight:
269.32
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

solid

color

white to off-white

mp

163 °C

solubility

H2O: soluble

storage temp.

2-8°C

SMILES string

[K+].CC(=O)Nc1ccc(OS([O-])(=O)=O)cc1

InChI

1S/C8H9NO5S.K/c1-6(10)9-7-2-4-8(5-3-7)14-15(11,12)13;/h2-5H,1H3,(H,9,10)(H,11,12,13);/q;+1/p-1

InChI key

AJYPYWFCUWHZMZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Paracetamol is an aminophenol derivative. It functions as an analgesic and antipyretic drug, which has weak inflammatory effect. Paracetamol is used to treat pyrexia and mild to moderate pain. It might inhibit atherosclerosis through its antioxidant activity.

Application

Paracetamol sulfate potassium salt has been used as a standard for HPLC assays of acetaminophen.

Biochem/physiol Actions

Phase II metabolite of paracetamol.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Paracetamol sulfate potassium salt is soluble in water.

Other Notes

Occasionally, it may be necessary to supply as the sodium salt

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paracetamol: past, present, and future.
Prescott LF
American Journal of Therapeutics, 7(2), 143-147 (2000)
D Bougie et al.
Blood, 97(12), 3846-3850 (2001-06-05)
In patients suspected of having drug-induced immune thrombocytopenia, antibodies reactive with normal platelets in the presence of the suspect drug can sometimes be identified, but negative results are often obtained. One reason for this is that drug metabolites, formed in
Karel Allegaert et al.
Acta paediatrica (Oslo, Norway : 1992), 94(9), 1273-1279 (2005-11-11)
Major changes in drug clearance and metabolism are observed during infancy, in part based on ontogenic regulation of various metabolic pathways. Since paracetamol provides a good substrate to study UGT (1A6) activity, urinary metabolites of propacetamol were determined in neonates
Hayati Filik et al.
Chemical & pharmaceutical bulletin, 54(6), 891-896 (2006-06-07)
In the present paper, conventional spectrophotometry in conjunction with cloud point extraction-preconcentration were investigated as alternative methods for paracetamol (PCT) assay in urine samples. Cloud point extraction (CPE) was employed for the preconcentration of p-aminophenol (PAP) prior to spectrophotometric determination
Caroline D van der Marel et al.
European journal of clinical pharmacology, 59(3), 243-251 (2003-05-23)
Data concerning metabolism of paracetamol in infants are scant. Previous studies have examined urinary metabolite recovery rates after a single dose of paracetamol in either neonates (<6 weeks) or children (3-9 years). There are no studies investigating infants. Infants (

Articles

Human epithelial intestinal colonic organoids can be used as an alternative to Caco-2 drug permeability assays for drug screening and compound toxicity testing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service