Skip to Content
Merck
All Photos(1)

Key Documents

U6881

Sigma-Aldrich

U-73343

Synonym(s):

1-[6-[((17β)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]-2,5-pyrrolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H42N2O3
CAS Number:
Molecular Weight:
466.66
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: soluble

SMILES string

[H][C@]12CC[C@]3(C)[C@H](CC[C@@]3([H])[C@]1([H])CCc4cc(OC)ccc24)NCCCCCCN5C(=O)CCC5=O

InChI

1S/C29H42N2O3/c1-29-16-15-23-22-10-8-21(34-2)19-20(22)7-9-24(23)25(29)11-12-26(29)30-17-5-3-4-6-18-31-27(32)13-14-28(31)33/h8,10,19,23-26,30H,3-7,9,11-18H2,1-2H3/t23-,24-,25+,26+,29+/m1/s1

InChI key

CJHWFIUASFBCKN-ZRJUGLEFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

U-73343 is used to assess the involvement of G protein-coupled 1-phosphatidylinositol-phosphodiesterase or phospholipase C in receptor-mediated cell activation. It functions downstream of phospholipase C to block receptor-mediated phospholipase D activation.

Application

U-73343 has been used for studying the role of phospholipase C (PLC) signaling in modulating epithelial Na+ channel (ENaC) function in gerbil Reissner′s membranes (RMs). U-73343 has also been used as a negative control for PLC inhibition in mechanically stimulated (MS) and its neighboring (NB) cells.
U-73343 has been used in measuring the fluorescence of intracellular Ca2+.

Biochem/physiol Actions

Inactive analog of U73122; used as a negative control.

Preparation Note

U-73343 is soluble in chloroform at 10 mg/ml and yields a clear, colorless to faint yellow solution. It is also soluble in ethanol (0.93 mg/ml) and DMSO (2.3 mg/ml), but is insoluble in water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuji Karashima et al.
Pflugers Archiv : European journal of physiology, 457(1), 77-89 (2008-05-08)
The transient receptor potential channel of the ankyrin-binding repeat subfamily, TRPA1, is a Ca(2+)-permeable non-selective cation channel that depolarizes the plasma membrane and causes Ca(2+) influx. A typical feature of TRPA1 is its rapid desensitization following activation by agonists such
Ayako Kataoka et al.
Journal of neurochemistry, 108(1), 115-125 (2008-11-19)
Microglia are implicated as a source of diverse proinflammatory factors in the CNS. Extracellular nucleotides are well known to be potent activators of glial cells and trigger the release of cytokines from microglia through purinergic receptors. However, little is known
R J Smith et al.
The Journal of pharmacology and experimental therapeutics, 253(2), 688-697 (1990-05-01)
1-[6-[[17 beta-3-Methoxyestra-1,3,5(10)-trien-17-yl]amino]hexyl]- 1H-pyrrole-2,5-dione (U-73122), an inhibitor of phospholipase C (PLC)-dependent processes in human platelets, was found to be a potent inhibitor of human polymorphonuclear neutrophil (PMN) activation by structurally unrelated receptor-specific agonists. U-73122 caused a time- and concentration-dependent (0.1-1 microM)
U73122 and U73343 inhibit receptor-mediated phospholipase D activation downstream of phospholipase C in CHO cells
Bosch RR, et al.
European Journal of Pharmacology, 346(2-3), 345-351 (1998)
Jue-Long Wang et al.
Toxicology letters, 160(1), 76-83 (2005-07-20)
The effect of the environmental toxicant nonylphenol on cytosolic free Ca2+ concentration ([Ca2+]i) and proliferation has not been explored in human osteoblast-like cells. This study examined whether nonylphenol alters Ca2+ levels and causes cell death in MG63 human osteosarcoma cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service