Skip to Content
Merck

U4010

Urokinase from human kidney cells

lyophilized powder

Synonym(s):

u-Plasminogen activator

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
232-917-9
NACRES:
NA.54
MDL number:
EC Number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

lyophilized powder

mol wt

heavy chain 30,400 Da, light chain 2000 Da

storage temp.

2-8°C

General description

Supplied as a lyophilized powder containing human albumin, mannitol and NaCl.
The principle active ingredient is the low molecular weight form of urokinase, and consists of an A chain of 2,000 daltons linked by a sulfhydyl bond to a B chain of 30,400 daltons.

Application

Urokinase from human kidney cells has been used as a component of digestion mix for three-dimensional fibrin culture. It has also been used in plasminogen activation assay.

Biochem/physiol Actions

Urokinase is a protease that cleaves plasminogen to form plasmin. It binds to cell surface receptors (uPAR) from which it regulates cell adhesion and migration during tissue remodeling and which also activate intracellular signaling pathways. Urokinase is composed of three domains: the amino terminal domain which is homologous to epidermal growth factor is attached through a central kringle domain to the protease domain in the carboxyl terminus.
Urokinase was shown to augment proliferation of renal tumor cells.

Other Notes

Note: Higher activities are obtained when human plasminogen is used as substrate.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library