Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.24
UNSPSC Code:
41121800
Pore size:
0.1 μm pore size
Filter capacity:
250 mL
Packaging:
case of 12 ea
Material:
polyethersulfone , polyethersulfone membrane
Manufacturer/tradename:
Nalgene 568-0010
material
polyethersulfone , polyethersulfone membrane
packaging
case of 12 ea
manufacturer/tradename
Nalgene 568-0010
filter capacity
250 mL
pore size
0.1 μm pore size
General description
Increase filtration efficiency by using the Nalgene® Rapid-Flow disposable filter units featuring the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination when filtering cell culture media, serum, additives and buffers. With a graduated upper chamber and Polyethersulfone (PES) membrane this filter unit offer higher aqueous flow rates; lower extractables (no wetting agent); lower protein binding; and greater strength, capacity, and chemical resistance than cellulose acetate, nitrocellulose, or nylon.
Application
Nalgene™ vacuum filtration system is suitable for the filtration of serum, cell culture medium, tissue culture, buffers and additives.
Features and Benefits
- Colour coded according to membrane material
- Devices feature the fastest flow rate and lowest clogging of comparable alternatives
- 0.1um PES available for mycoplasma protection
- 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
- Non-pyrogenic and endotoxin free
- Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit
- Graduated Upper chamber
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Explore all of our products under Nalgene® vacuum filtration system
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.